Skip to Content
Merck

Skip To

P0106

Pefloxacin mesylate dihydrate

Synonym(s):

3-Quinolinecarboxylic acid, 1-ethyl-6-fluoro-1,4-dihydro-7-(4-methyl-1-piperazinyl)-4-oxo-, monomethanesulfonate, dihydrate, Pefloxacine monomethanesulfonate dihydrate, Pefloxacinium methanesulfonate dihydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C17H20FN3O3 • CH4O3S • 2H2O
CAS Number:
Molecular Weight:
465.49
NACRES:
NA.76
PubChem Substance ID:
UNSPSC Code:
51102829
EC Number:
274-613-9
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Segment

solubility

H2O: 50 mg/mL

antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria

mode of action

DNA synthesis | interferes, enzyme | inhibits

storage temp.

2-8°C

SMILES string

O.O.CS(O)(=O)=O.CCN1C=C(C(O)=O)C(=O)c2cc(F)c(cc12)N3CCN(C)CC3

InChI

1S/C17H20FN3O3.CH4O3S.2H2O/c1-3-20-10-12(17(23)24)16(22)11-8-13(18)15(9-14(11)20)21-6-4-19(2)5-7-21;1-5(2,3)4;;/h8-10H,3-7H2,1-2H3,(H,23,24);1H3,(H,2,3,4);2*1H2

InChI key

LEULAXMUNMRLPW-UHFFFAOYSA-N

General description

Chemical structure: fluoroquinolone

Application

Pefloxacin is a synthetic broad-spectrum fluoroquinolone antibiotic that is effective against most gram-negative and gram-positive bacteria. It is used to treat gonococcal urethritis and for gram-negative-bacterial infections in the gastrointestinal system and the genitourinary tract. Pefloxacin mesylate dihydrate has been used to induce achilles tendon toxicity in rodents[1]. It′s cytotoxicity and uptake have been studied in primary cultures of rat hepatocytes[2].

Biochem/physiol Actions

Pefloxacin inhibits the bacterial enzymes DNA gyrase and topoisomerase IV, which are needed for the transcription and replication of bacterial DNA. DNA gyrase is thought to be the primary quinolone target for gram-negative bacteria. Topoisomerase IV is thought to be the primary target in gram-positive organisms. The inhibition of the topoisomerases results in strand breakage of the bacterial chromosome, supercoiling, and resealing. Therefore, DNA replication and transcription is inhibited . Pefloxacin is an analog of norfloxacin.
Pefloxacin is a synthetic fluoroquinolone that functions an antibacterial agent. It is an analog of norfloxacin.
Mode of Action: Pefloxacin prevents bacterial DNA replication by inhibiting DNA gyrase.
Antimicrobial spectrum: Pefloxacin is highly active against Staphylococcus aureus, E. coli, other enterobacteria, and Pseudomonas aeruginosa. Active against gram-positive bacteria and excellent activity against gram-negative bacteria.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters