Skip to Content
Merck

O3639

Ondansetron hydrochloride dihydrate

≥98% (HPLC), powder, 5-HT₃serotonin receptor antagonist

Synonym(s):

1,2,3,9-Tetrahydro-9-methyl-3-[(2-methyl-1H-imidazol-1-yl)methyl]-4H-carbazol-4-one hydrochloride, GR 38032F

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H19N3O · HCl · 2H2O
CAS Number:
Molecular Weight:
365.85
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Ondansetron hydrochloride dihydrate, ≥98% (HPLC), powder

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white

solubility

H2O: >5 mg/mL

storage temp.

−20°C

SMILES string

Cl[H].[H]O[H].[H]O[H].Cc1nccn1CC2CCc3c(C2=O)c4ccccc4n3C

InChI

1S/C18H19N3O.ClH.2H2O/c1-12-19-9-10-21(12)11-13-7-8-16-17(18(13)22)14-5-3-4-6-15(14)20(16)2;;;/h3-6,9-10,13H,7-8,11H2,1-2H3;1H;2*1H2

InChI key

VRSLTNZJOUZKLX-UHFFFAOYSA-N

Gene Information

human ... HTR3A(3359)

Application

Ondansetron hydrochloride dihydrate has been used as a 5-HT3 antagonist to attenuate rostro ventromedial medulla (RVM)/cholecystokinin (CCK)-induced mechanical hypersensitivity and to block the effects of trichostatin A (TSA) on 5-HT3 protein.

Biochem/physiol Actions

Ondansetron, (3RS)-9-methyl-3-[(2-methyl-1H-imidazol-1-yl) methyl] 1,2,3,9-tetrahydro-4H-carbazol-4-one hydrochloride dehydrate acts as a selective 5-hydroxytryptamine type 3 (5-HT3) serotonin receptor antagonist. It is used as an antiemetic to control nausea and vomiting associated with chemotherapy and radiotherapy.
5-HT3 serotonin receptor antagonist

Features and Benefits

This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.


Still not finding the right product?


signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library