Skip to Content
Merck
All Photos(1)

Key Documents

O1385

Sigma-Aldrich

Ozagrel hydrochloride hydrate

≥98% (HPLC), solid

Synonym(s):

(E)-3-[4-(Imidazol-1-ylmethyl)phenyl]propenoic acid hydrochloride hydrate, OKY-046

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H12N2O2 · HCl · xH2O
CAS Number:
Molecular Weight:
264.71 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

storage condition

desiccated

color

white to off-white

solubility

H2O: soluble

storage temp.

room temp

SMILES string

O.Cl.OC(=O)\C=C\c1ccc(Cn2ccnc2)cc1

InChI

1S/C13H12N2O2.ClH.H2O/c16-13(17)6-5-11-1-3-12(4-2-11)9-15-8-7-14-10-15;;/h1-8,10H,9H2,(H,16,17);1H;1H2/b6-5+;;

InChI key

OWIZTYOMGVTSDP-TXOOBNKBSA-N

Application

Ozagrel hydrochloride hydrate has been used as a thromboxane synthase inhibitor:
  • in high cholesterol-diet rats (HC) to test its effect on arteriolar constriction
  • to pre-treat mice to test its effect on lipopolysaccharide (LPS)-induced behavioral changes
  • to test its effect on arteriolar vasoconstriction in hyperglycemic mice

Biochem/physiol Actions

Ozagrel is a selective thromboxane A2 synthase (TXA2) inhibitor.
Ozagrel is useful in treating lacunar and thrombotic stroke. It blocks vasoconstriction and platelet aggregation. Ozagrel aids in improving the prostacyclin (PGI2)/ thromboxane A2 (TXA2) in the acute phase of cerebral ischemia. It improves endothelial dysfunction in L-methionine-induced hyperhomocysteinemia (HHcy)-induced vascular cognitive impairment and dementia (VCID).

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seungjun Lee et al.
Microvascular research, 76(3), 217-223 (2008-08-23)
Retinal blood flow decreases early in the progression of diabetic retinopathy; however, the mediators and mechanisms responsible for this decrease have yet to be determined. In this study, diabetes was induced by streptozotocin in rats, and retinal blood flow was
Yukito Shinohara et al.
Cerebrovascular diseases (Basel, Switzerland), 27(5), 485-492 (2009-03-27)
Edaravone, a free radical scavenger approved by the Japanese Ministry of Health, Labor and Welfare in 2001 for treating acute ischemic stroke, was recommended by the Japanese Guidelines for the Management of Stroke 2004. While edaravone also has a neuroprotective
Yuichi Abe et al.
Gastroenterology, 145(3), 658-667 (2013-05-21)
A 3-dimensional (3D) culture system for immortalized human hepatocytes (HuS-E/2 cells) recently was shown to support the lifecycle of blood-borne hepatitis C virus (HCV). We used this system to identify proteins that are active during the HCV lifecycle under 3D
K Shinagawa et al.
British journal of pharmacology, 131(2), 266-270 (2000-09-19)
1. The purpose of this study was to investigate the involvement of thromboxane A(2) (TXA(2)) in the cough response in a guinea-pig cough model. Here, we describe results obtained using a selective TXA(2) synthetase inhibitor, ozagrel, and a selective TXA(2)
Yukiko Enomoto et al.
Atherosclerosis, 209(2), 415-421 (2009-11-26)
The plasma level of soluble CD40 ligand (sCD40L), which induces pro-inflammatory and pro-atherogenic responses, is known to be elevated in atherosclerotic patients. In this study, we investigated the mechanism of sCD40L release from human platelets, focusing on the involvement of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service