Skip to Content
Merck
All Photos(2)

Key Documents

N9528

Sigma-Aldrich

Anti-Neuropeptide Y (NPY) antibody produced in rabbit

whole antiserum

Synonym(s):

NPY Antibody - Anti-Neuropeptide Y (NPY) antibody produced in rabbit, Npy Antibody

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

contains

15 mM sodium azide

species reactivity

rat, sheep, pig, human

packaging

antibody small pack of 25 μL

technique(s)

: suitable using suitable for IHC on rat brain perfused with 4% paraformaldehyde
indirect ELISA: 1:5,000-1:10,000 using using peptide coating at 0.5-1 μg/well in 0.05 M carbonate/bicarbonate buffer, pH 9.6, and pNPP as substrate

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

rat ... Npy(24604)

General description

Anti-Neuropeptide Y (NPY) is developed in rabbits using synthetic neuropeptide Y (porcine) conjugated to keyhole limpet hemocyanin (KLH) as the immunogen.
Neuropeptide Y (NPY), a 36-amino acid peptide amide, is a major regulatory neuropeptide widely distributed in the mammalian central (CNS) and peripheral nervous systems (PNS). NPY belongs to the pancreatic polypeptide family of peptides which are characterized by a common tertiary structure. Within this family, an intestinal peptide hormone, peptide YY (PYY), is most closely related to NPY. In the CNS, NPY is involved in regulation of blood pressure, memory processing, circadian rhythm, and stimulation of food intake. In the PNS, NPY has potent vasoconstrictor activity and acts as a neurotransmitter/neuromodulator of sympathetic neurons and adrenal glands. NPY is one of the most abundant peptides found in the CNS, widely distributed in the brain. High levels of NPY are present in the cerebral cortex, amygdaloid nuclei, hippocampal formation, and hypothalamus. In the PNS, NPY is found mainly in sympathetic neurons that innervate vascular smooth muscle, heart, and urogenital tract. The biological actions of NPY in the brain and periphery are mediated by at least two different NPY receptors, designated Y1 and Y2 receptor subtypes.

Specificity

The antibody recognizes by ELISA, NPY (human), NPY (porcine), NPY (sheep), NPY (13-36), NPY (18-36), and cross-reacts with PYY (human). It does not cross-react with neurokinin B (NKB), neurokinin A (NKA), substance P (SP), calcitonin, somatostatin and BSA.

Immunogen

synthetic neuropeptide Y (NPY, porcine), conjugated to KLH. The corresponding sequence is highly conserved (single amino acid substitution) in human and rat NPY and is highly conserved (84% identity) in the C-terminus region (24-36) of human peptide YY.

Application

Anti-Neuropeptide Y (NPY) antibody produced in rabbit has been used in:
  • single-labeling immunocytochemistry
  • immunofluorescence
  • immunostaining
  • dot blot
  • radioimmunoassay (RIA)
  • enzyme-linked immunosorbent assay (ELISA)

Biochem/physiol Actions

Neuropeptide Y (NPY) plays a vital role in feeding behaviour, anxiety, epilepsy, circadian, rhythm memory processing, sleep, pain and drug addiction. In addition, it is also implicated in regulation of appetite, body weight and obesity. It controls central endocrine secretion and has potent vasoactive effects on the cardiovascular system. NPY is involved in the pathogenesis of atherosclerosis and coronary artery disease.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jadwiga Konieczna et al.
PloS one, 8(11), e81906-e81906 (2013-12-07)
A poor prenatal environment brings about perturbations in leptin surge and hypothalamic circuitry that program impaired ability to regulate energy homeostasis in adulthood. Here, using a rat model of moderate maternal caloric restriction during gestation, we aimed to investigate whether
Nerve outgrowth and neuropeptide expression during the remodeling of human burn wound scars: A 7-month follow-up study of 22 patients
Altun V, et al.
Burns : Journal of the International Society For Burn Injuries, 27(7), 717-722 (2001)
Zuzanna Michalak et al.
Journal of neuropathology and experimental neurology, 71(9), 826-838 (2012-08-11)
Focal epilepsies are often associated with blood-brain barrier disruption. In 4 entorhinal cortex tissue samples and 13 hippocampal samples from patients with pharmacoresistent temporal lobe epilepsy, we observed immunoglobulin G (IgG) leakage in the parenchyma and IgG-positive neurons that had
Effects of photoperiod on estrogen receptor, tyrosine hydroxylase, neuropeptide Y, and beta-endorphin immunoreactivity in the ewe hypothalamus
Skinner DC and Herbison AE
Endocrinology, 138(6), 2585-2595 (1997)
Nora López et al.
Nutrients, 10(1) (2018-01-13)
Early nutrition plays an important role in development and may constitute a relevant contributor to the onset of obesity in adulthood. The aim of this study was to evaluate the long-term impact of maternal leucine (Leu) supplementation during lactation on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service