Skip to Content

N8513

L-Norleucine

suitable for amino acid analysis, BioReagent

Synonym(s):

(S)-(+)-2-Aminohexanoic acid, (S)-2-Aminocaproic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3(CH2)3CH(NH2)CO2H
CAS Number:
Molecular Weight:
131.17
NACRES:
NA.26
PubChem Substance ID:
eCl@ss:
32160406
UNSPSC Code:
12352209
EC Number:
206-321-4
MDL number:
Beilstein/REAXYS Number:
1721750
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

L-Norleucine, suitable for amino acid analysis, BioReagent

product line

BioReagent

Quality Segment

form

powder or crystals

color

white

mp

>300 °C (lit.)

suitability

suitable for amino acid analysis

application(s)

detection

SMILES string

CCCC[C@H](N)C(O)=O

InChI

1S/C6H13NO2/c1-2-3-4-5(7)6(8)9/h5H,2-4,7H2,1H3,(H,8,9)/t5-/m0/s1

InChI key

LRQKBLKVPFOOQJ-YFKPBYRVSA-N

Application

Highly purified internal standard for all amino acid analysis methods.

Other Notes

Non-essential amino acid.


Still not finding the right product?

Explore all of our products under L-Norleucine


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Skin Sens. 1

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library