Skip to Content
Merck

Skip To

N3786

α(2→3,6,8,9) Neuraminidase from Arthrobacter ureafaciens

Proteomics Grade, suitable for MALDI-TOF MS

Synonym(s):

Neuraminidase from Arthrobacter ureafaciens, Acyl-neuraminyl Hydrolase, Sialidase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.32
EC Number:
232-624-6
MDL number:
EC Number:
Specific activity:
≥25 U/vial
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

lyophilized powder

Quality Segment

quality

Proteomics Grade

specific activity

≥25 U/vial

suitability

suitable for MALDI-TOF MS

storage temp.

2-8°C

Application

Neuraminidase is an important deglycosylation enzyme capable of cleaving all non-reducing unbranched N-acetylneuraminic and N-glycolylneuraminic acid residues by hydrolysis of α(2→6), α(2→3), α(2→8), and α(2→9) linkages (affinity in the order given). Branched sialic acids may also be cleaved with the use of high concentrations of enzyme and prolonged incubations. Desialylated glycoproteins may then be further characterized by treatment with various exoglycosidases resulting in partial or complete O-deglycosylation. SDS-PAGE and MALDI-TOF MS are typically utilized in purification, structural analysis, and sequencing process. These techniques also remove heterogeneity and charge from the glycoprotein.

Packaging

Provided with 5× concentrate reaction buffer.

Physical form

Lyophilized powder

Analysis Note

The buffer solution has been specifically formulated to assure maximum enzyme stability and assay sensitivity.

Other Notes

One unit will release 1 nmole of 4-methylumbelliferone from 2-(4-methylumbelliferyl) α-D-N-acetylneuraminic acid per minute at pH 5.5 at 37° C.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
N7885N6514N3001
specific activity

≥25 U/vial

specific activity

1-5 units/mg protein (Lowry, using NAN-lactose)

specific activity

8-24 units/mg protein (Lowry, using NAN-lactose)

specific activity

≥2 units/mg protein (mucin), ≥6 units/mg protein (using 4MU-NANA)

form

lyophilized powder

form

buffered aqueous solution

form

buffered aqueous solution

form

lyophilized powder

suitability

suitable for MALDI-TOF MS

suitability

-

suitability

-

suitability

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

Quality Level

200

Quality Level

300

Quality Level

200

Quality Level

200

quality

Proteomics Grade

quality

-

quality

-

quality

-


Still not finding the right product?


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • Neuraminidase 5X Reaction Buffer 1.5 mL
    SDS

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters