Skip to Content
Merck

Skip To

N149

Nimodipine

≥98% (HPLC), L-type Ca2+ channel antagonist, powder

Synonym(s):

1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic acid 2-methoxyethyl 1-methylethyl ester, Isopropyl 2-methoxyethyl 1,4-dihydro-2,6-dimethyl-4-(m-nitrophenyl)-3,5-pyridinedicarboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C21H26N2O7
CAS Number:
Molecular Weight:
418.44
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
EC Number:
266-127-0
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Nimodipine,

assay

≥98% (HPLC)

Quality Segment

form

powder

color

off-white to yellow

solubility

methanol: 62.5 mg/mL, 45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: insoluble, H2O: insoluble

originator

Bayer

SMILES string

COCCOC(=O)C1=C(C)NC(C)=C(C1c2cccc(c2)[N+]([O-])=O)C(=O)OC(C)C

InChI

1S/C21H26N2O7/c1-12(2)30-21(25)18-14(4)22-13(3)17(20(24)29-10-9-28-5)19(18)15-7-6-8-16(11-15)23(26)27/h6-8,11-12,19,22H,9-10H2,1-5H3

InChI key

UIAGMCDKSXEBJQ-UHFFFAOYSA-N

Application

Nimodipine has been used:
  • as a L-type calcium channel (LTCC) inhibitor, to evaluate its neuroprotective activity in the vibrosections
  • as a standard, in the enantioseparation of chiral drugs by high performance liquid chromatography (HPLC)
  • in the pharmacological studies for the measurement of spine voltage escape

Biochem/physiol Actions

Nimodipine is a potent L-type Ca2+ channel antagonist.
Nimodipine enhances the survival of dopaminergic substantia nigra neurons.

Features and Benefits

This compound is featured on the Calcium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Bayer. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Disclaimer

Photosensitive

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
482200BP651PHR1293
Nimodipine British Pharmacopoeia (BP) Reference Standard

BP651

Nimodipine

description

-

description

An L-type Ca2+ channel blocker.

description

British Pharmacopoeia (BP) Reference Standard

description

Pharmaceutical Secondary Standard; Certified Reference Material

assay

≥98% (HPLC)

assay

>98% (HPLC)

assay

-

assay

-

form

powder

form

solid

form

solid

form

-

Quality Level

200

Quality Level

100

Quality Level

-

Quality Level

300

solubility

methanol: 62.5 mg/mL, H2O: insoluble, 45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: insoluble

solubility

DMSO: 25 mg/mL, methanol: soluble

solubility

-

solubility

-

color

off-white to yellow

color

yellow

color

-

color

-

Gene Information

human ... ADORA3(140), CACNA1C(775), CACNA1D(776), CACNA1F(778), CACNA1S(779), CACNG1(786), NR3C2(4306)
rat ... Adora1(29290), Adora2a(25369)

Gene Information

-

Gene Information

human ... CACNA1C(775), CACNA1D(776), CACNA1F(778), CACNA1S(779), NR3C2(4306)

Gene Information

human ... CACNA1C(775), CACNA1D(776), CACNA1F(778), CACNA1S(779), NR3C2(4306)


Still not finding the right product?

Explore all of our products under Nimodipine


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters