Skip to Content
Merck
All Photos(1)

Key Documents

N0258

Sigma-Aldrich

N-Nitrosodiethylamine

ISOPAC®

Synonym(s):

Diethylnitrosamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NNO
CAS Number:
Molecular Weight:
102.14
Beilstein:
1744991
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.25

Quality Level

form

liquid

bp

177 °C (lit.)

density

0.95 g/mL (lit.)

SMILES string

CCN(CC)N=O

InChI

1S/C4H10N2O/c1-3-6(4-2)5-7/h3-4H2,1-2H3

InChI key

WBNQDOYYEUMPFS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-Nitrosodiethylamine (NDEA) has been used as a hepatocarcinogen/DNA damaging agent. It has been used as a calibration standard for gas–liquid chromatography.

Biochem/physiol Actions

N-Nitrosodiethylamine (NDEA) is a potent hepatocarcinogenic dialkylnitrosoamine. It is mainly present in tobacco smoke, water, cheddar cheese, cured, fried meals and many alcoholic beverages. NDEA is responsible for the changes in the nuclear enzymes associated with DNA repair/replication. It results in various tumors in all animal species. The main target organs are the nasal cavity, trachea, lung, esophagus and liver.

Packaging

Packaged in a 100 mL serum bottle with silicon/PTFE septum and aluminum tear seal.

Caution

Injecting any compatible solvent permits preparation of any desired strength solution without exposure.

Reconstitution

Dissolving the contents in 100 mL of solvent yields a 1% solution.

Legal Information

Isopac is a registered trademark of Merck KGaA, Darmstadt, Germany

ISOPAC of

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Chemopreventive effect of hydroethanolic extract of Euphorbia neriifolia leaves against DENA-induced renal carcinogenesis in mice.
Pracheta P
Asian Pacific Journal of Cancer Prevention, 12, 677-677 (2011)
Proteomic profiling of expression of proteasomal subunits from livers of mice treated with diethylnitrosamine.
Yuan F
Proteomics, 13, 389-389 (2013)
The detection of DNA adducts (risk factors for DNA damage). A method for genomic DNA, the results and some effects of nutritional intervention.
Howard JM
Journal of Nutritional & Environmental Medicine, 12, 19-31 (2002)
Yewei Liu et al.
International journal of cancer, 143(7), 1817-1828 (2018-05-01)
To better inform the next clinical trials of vascular disrupting agent combretastatin-A4-phosphate (CA4P) in patients with hepatic malignancies, this preclinical study aimed at evaluating CA4P therapeutic efficacy in rats with primary hepatocellular carcinomas (HCCs) of a full spectrum of differentiation
Dongyao Wang et al.
EBioMedicine, 46, 119-132 (2019-08-04)
Patients undergoing surgical resection of hepatocellular carcinoma (HCC) are at risk of recurrence; however, the underlying mechanism remains poorly understood. Through the analysis of gene expression profiles in tumour and matched normal tissues from patients with hepatocellular carcinoma (HCC), we

Articles

DNA damage and repair mechanism is vital for maintaining DNA integrity. Damage to cellular DNA is involved in mutagenesis, the development of cancer among others.

Cancer research has revealed that the classical model of carcinogenesis, a three step process consisting of initiation, promotion, and progression, is not complete.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service