Skip to Content

N0163

Nitrate Reductase from Arabidopsis thaliana

vial of ≥0.5 unit

Synonym(s):

NADH:nitrate oxidoreductase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352204
NACRES:
NA.54
EC Number:
MDL number:
Recombinant:
expressed in Pichia pastoris
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in Pichia pastoris

Quality Segment

form

lyophilized powder

packaging

vial of ≥0.5 unit

shipped in

wet ice

storage temp.

−20°C

Application

Catalyzes the NADH-dependent reduction of nitrate to nitrite.
Nitrate reductase from Arabidopsis thaliana has been used in a study to assess the amino acid sequence of chicken hepatic sulfite oxidase.

Biochem/physiol Actions

Nitrate reductase activity is induced in Arabidopsis thaliana plants by sumoylation via the E3 ligase activity of AtSIZ1.

Physical form

Supplied as a lyophilized powder containing 50 mM MOPS, pH 7.0, 0.1 mM EDTA and a proprietary sugar

Other Notes

One unit will reduce 1.0 micromole of nitrate to nitrite per min in a NADH system at pH 7.5 at 30 deg C.


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library