Skip to Content
Merck

N0128

Nitrilotriacetic acid disodium salt

Sigma Grade, ≥99%

Synonym(s):

N,N-Bis(carboxymethyl)glycine, NTA, Triglycollamic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H7NO6Na2
CAS Number:
Molecular Weight:
235.10
NACRES:
NA.22
PubChem Substance ID:
UNSPSC Code:
12352100
EC Number:
239-484-5
MDL number:
Assay:
≥99%
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Sigma Grade

Quality Segment

assay

≥99%

form

powder

SMILES string

[Na+].[Na+].OC(=O)CN(CC([O-])=O)CC([O-])=O

InChI

1S/C6H9NO6.2Na/c8-4(9)1-7(2-5(10)11)3-6(12)13;;/h1-3H2,(H,8,9)(H,10,11)(H,12,13);;/q;2*+1/p-2

InChI key

RHPXYIKALIRNFA-UHFFFAOYSA-L

Application

An excellent chelating agent.

Disclaimer

Possibly carcinogenic.


pictograms

Health hazardExclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2

Storage Class

11 - Combustible Solids

wgk

WGK 2

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library