Skip to Content
Merck

Skip To

ML0013

Glycolysis/Gluconeogenesis Metabolite Library

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


application(s)

advanced drug delivery

storage temp.

−20°C

Other Notes

Contains 10 mg each of the glycolysis and gluconeogenesis pathway metabolite standards packaged individually.
For additional information on our range of Biochemicals, please complete this form.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
ML0011ML0012ML0010
description

-

description

-

description

-

description

-

application(s)

advanced drug delivery

application(s)

-

application(s)

advanced drug delivery

application(s)

advanced drug delivery

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • α-D-Glucose, anhydrous, 96% 15 mg
    SDS

  • Biotin, ≥99% (HPLC), lyophilized powder 10 mg
    SDS

  • Dihydroxyacetone phosphate dilithium salt, ≥93% dry basis (enzymatic) 10 mg
    SDS

  • D-Fructose 6-phosphate disodium salt hydrate, ≥98%, amorphous powder 10 mg
    SDS

  • D-Fructose 1,6-bisphosphate trisodium salt hydrate, ≥98% (TLC) 10 mg
    SDS

  • α-D-Glucose 1-phosphate dipotassium salt hydrate, ≥99% (HPLC), BioXtra 10 mg
    SDS

  • Guanosine 5′-diphosphate sodium salt, Type I, ≥96% (HPLC) 10 mg
    SDS

See All (17)

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1C - Skin Sens. 1 - STOT SE 2 - STOT SE 3

target_organs

Eyes,Central nervous system, Respiratory system

supp_hazards

Storage Class

8A - Combustible corrosive hazardous materials

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters