Skip to Content
Merck
All Photos(2)

Key Documents

MDQ1

Sigma-Aldrich

Imprint® Methylated DNA Quantification Kit

To measure global DNA methylation shifts from as low as 10 ng DNA

Synonym(s):

DNA Methylation Quantification, DNA methylation shift

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.54

General description

Imprint™ Methylated DNA Quantification technology provides a rapid and reliable method to measure global DNA methylation shifts. The Imprint Methylated DNA Quantification Kit contains all reagents required to detect relative levels of methylated DNA. Up to 200 ng of purified DNA is bound to the wells of the assay strip. The methylated DNA is detected using the Capture and Detection antibodies, then quantified colorimetrically. The amount of methylated DNA present in the sample is proportional to the absorbance measured.

Application

Imprint® Methylated DNA Quantification Kit has been used in global DNA methylation analysis.

Features and Benefits

  • ELISA based format
  • Procedure completes in 4 easy to follow steps
  • All reagents supplied including control methylated DNA
  • Detection limit is 5 ng of fully methylated DNA
  • Input DNA may be as low as 10-200 ng
  • Procedure completes in 3.5 hours
  • 96 well format allows option of studying single samples or high-throughput studies
  • Optimized antibody and reagents with high specificity to 5-mC; no cross-reactivity to unmethylated cytosine
  • May be used with adherent and suspension cells, tissues, plasma and other body fluids such as saliva, urine and serum

Legal Information

Imprint is a registered trademark of Merck KGaA, Darmstadt, Germany

Kit Components Also Available Separately

Product No.
Description
SDS

  • M8570Methylated Control DNA, 50 ng/μLSDS

Pictograms

CorrosionExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Met. Corr. 1 - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Felicitas Thol et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 29(21), 2889-2896 (2011-06-15)
To study the incidence and prognostic impact of mutations in DNA methyltransferase 3A (DNMT3A) in patients with acute myeloid leukemia. A total of 489 patients with AML were examined for mutations in DNMT3A by direct sequencing. The prognostic impact of
Alok Mishra et al.
Methods in molecular biology (Clifton, N.J.), 863, 15-31 (2012-02-24)
Epigenetics is an emerging science that can help to explain carcinogenesis. The possibility that carcinogenesis may originate in a stem cell process was proposed recently. Stem cells are generated and contribute to tumor formation during the process of tumor development.
Roman Thaler et al.
The British journal of nutrition, 101(5), 743-749 (2008-08-08)
The impact of nutrition on the epigenetic machinery has increasingly attracted interest. The aim of the present study was to demonstrate the effects of various diets on methylation and gene expression. The antioxidative enzyme mitochondrial superoxide dismutase (MnSOD) was chosen
Mounira Krifa et al.
Journal of experimental & clinical cancer research : CR, 32, 30-30 (2013-05-22)
Several reports have described the potential effects of natural compounds as anti-cancer agents in vitro as well as in vivo. The aim of this study was to evaluate the anti-cancer effect of Limoniastrum guyonianum aqueous gall extract (G extract) and
Rauf Ahmad Najar et al.
The Journal of nutritional biochemistry, 53, 121-132 (2017-12-09)
The present study has been designed to determine the effect of folate modulation (deficiency/supplementation) with aging on the promoter methylation of tumor suppressor and proto-oncogenes to understand the underlying mechanism of epigenetic alterations. Folate deficiency was induced for 3 and

Articles

Global DNA Modification Kit (MDQ1) monitors DNA methylation via sandwich ELISA-like assay, aiding in epigenetic research.

Global DNA Modification Kit (MDQ1) monitors DNA methylation via sandwich ELISA-like assay, aiding in epigenetic research.

Global DNA Modification Kit (MDQ1) monitors DNA methylation via sandwich ELISA-like assay, aiding in epigenetic research.

Global DNA Modification Kit (MDQ1) monitors DNA methylation via sandwich ELISA-like assay, aiding in epigenetic research.

Protocols

The Imprint® Methylated DNA Quantification Kit provides a high-throughput, non-radioactive means of measuring global DNA methylation shifts.

Chromatin Immunoprecipitation qPCR for studying gene regulation across conditions.

Chromatin Immunoprecipitation qPCR for studying gene regulation across conditions.

Chromatin Immunoprecipitation qPCR for studying gene regulation across conditions.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service