Skip to Content
Merck

MAK485

Pyruvate Kinase Assay Kit

sufficient for 100 colorimetric or fluorometric tests

Synonym(s):

PK Activity Assay Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.84
UNSPSC Code:
12352204
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

application(s)

pharmaceutical

detection method

colorimetric, fluorometric

relevant disease(s)

metabolic disorders; endocrinological disorders; cancer; hematological disorder

shipped in

wet ice

storage temp.

−20°C

General description

Pyruvate Kinase (PK) is an enzyme involved in glycolysis. It catalyzes the transfer of a phosphate group from phosphoenolpyruvate (PEP) to ADP, yielding one molecule of pyruvate and one molecule of ATP. Pyruvate kinase deficiency, a genetic disease, is caused by a lack of pyruvate kinase and slows down the process of glycolysis. Pyruvate kinase is also involved in gluconeogenesis, a biochemical pathway in which the liver generates glucose from pyruvate and other substrates.

Application

The kit is suitable for pyruvate kinase activity determination in plasma, serum, and tissue samples, as well as for studying the effects of drugs on pyruvate kinase activity.

Biochem/physiol Actions

The Pyruvate Kinase Assay Kit provides a simple, direct and automation-ready procedure for measuring pyruvate kinase activity. In this assay PEP and ADP are catalyzed by pyruvate kinase to generate pyruvate and ATP. The color intensity of the reaction product at 570 nm or fluorescence intensity at λEx = 530 nm/λEm = 590 nm is directly proportional to the pyruvate generated by the pyruvate kinase in the sample.

Features and Benefits

  • 0.1 to 50 U/L for colorimetric assay and 0.01 to 2 U/L for fluorometric assay



Storage Class

10 - Combustible liquids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library