Skip to Content
Merck
All Photos(3)

Documents

MAK174

Sigma-Aldrich

Fatty Acid Extraction Kit, Low Standard

greener alternative

sufficient for 40 extractions

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.25

usage

sufficient for 40 extractions

greener alternative product characteristics

Waste Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

application(s)

cosmetics
food and beverages
sample preservation

greener alternative category

relevant disease(s)

cancer

storage temp.

room temp

General description

Lipids are structural components of cell membranes that play a critical role in gene transcription, signaling, and metabolism. Several lipid species exist in biological systems, including phospholipids, triglycerides, and fatty acids. When extracting fatty acids at low or very low levels, such as in cellular matrices, use the MAK174 fatty acid extraction kit. When extracting fatty acids at normal or high levels, such as in food matrices, use the MAK338 fatty acid extraction kit.
The Folch method has been conventionally used to extract lipids containing fatty acids from biological samples, using chloroform, methanol, and water to separate lipids from aqueous-soluble compounds. In this procedure, lipids are retained in the lower chloroform layer; whereas, aqueous-soluble compounds are retained in the upper methanol-water layer. The sample is then centrifuged to achieve uniform separation and the bottom chloroform layer is transferred with a pipette to a new test tube.

Application

Fatty Acid Extraction Kit has been used for fatty acid extraction.

Suitability

This kit is suitable for the extraction of lipids from a variety of samples such as whole blood, serum, biological tissue, food samples, and oil.

Principle

The Fatty Acid Extraction Kit shortens this extraction process by eliminating the need to prepare solvents and standards, centrifugation, and pipetting. Once the sample is homogenized and dissolved in the Extraction Solvent supplied in our kit, it is inverted twice and poured into the syringe containing a filter, which preferentially elutes the chloroform layer containing total lipids. A portion of the total lipid extract containing fatty acids can then be transesterified for GC-FID analysis.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product reduces solvent waste over traditional methods. Find details here.

Other Notes

Experts have tested the Fatty Acid Extraction Kit. Watch the Video to learn more and to see how the technology works.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 1 - STOT SE 3

Target Organs

Central nervous system, Eyes, Liver,Kidney

Storage Class Code

3 - Flammable liquids

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service