Skip to Content
Merck

MAK152

Colorimetric Sphingomyelinase Assay Kit

sufficient for 200 colorimetric tests

Synonym(s):

Neutral Sphingomyelinase Assay Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.84
UNSPSC Code:
12161503
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


usage

sufficient for 200 colorimetric tests

detection method

colorimetric

relevant disease(s)

rheumatological diseases; cancer

storage temp.

−20°C

General description

Sphingomyelinase (SMase), or sphingomyelin phosphodiesterase, catalyzes the hydrolytic cleavage of sphingolipid to phosphocholine and ceramide, a second messenger involved in multiple cellular responses including the cellular response to stress. There are at least 5 types of SMases identified which are classified according to their cation dependence, cellular location, and pH optima for activity and include the acidic and neutral sphingomyelinase. Neutral SMases, along with the acidic SMases, are thought to be major contributors to the production of ceramide in response to cellular stress.

Application

This kit is suitable for the detection of sphingomyelinase activity in a variety of samples such as blood and cell extracts or for screening sphingomyelinase inhibitors and activators.

Biochem/physiol Actions

The Sphingomyelinase Assay Kit provides a simple and direct procedure for measuring neutral SMase activity in a variety of samples such as blood or cell extracts, or for screening SMase activators and inhibitors. SMase activity is determined by a coupled enzyme assay, which results in a colorimetric (655 nm) product proportional to the SMase activity present.

Features and Benefits

Compatible with high-throughput handling systems.


signalword

Danger

Storage Class

10 - Combustible liquids

wgk

WGK 3

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - ED ENV 1 - Eye Dam. 1 - Resp. Sens. 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library