Skip to Content
Merck

MAK142

Fluorometric Intracellular Ros Kit

sufficient for 200 fluorometric tests (Deep red)

Synonym(s):

Intracellular ROS Fluorescence Assay

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12161503
EC Number:
200-664-3
NACRES:
NA.25
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


usage

sufficient for 200 fluorometric tests (Deep red)

application(s)

cosmetics
food and beverages

detection method

fluorometric

relevant disease(s)

neurological disorders; cancer

storage temp.

−20°C

General description

Reactive oxygen species (ROS) are generated as a result of the reduction of oxygen during aerobic respiration and by various enzymatic systems within the cell. At physiological levels, ROS contribute to cell signaling and host defense. Increased ROS generation, above the detoxification capacity of the biological system, results in oxidative stress and cellular damage. The main damage to cells results from the ROS-induced alteration of macromolecules such as polyunsaturated fatty acids in membrane lipids, essential proteins, and DNA. ROS has been implicated in disease states, such as Alzheimer′s disease, Parkinson′s disease, cancer, and aging.

Application


  • Diethyl Blechnic Exhibits Anti-Inflammatory and Antioxidative Activity via the TLR4/MyD88 Signaling Pathway in LPS-Stimulated RAW264.7 Cells.: This article uses fluorometric methods to explore the anti-inflammatory and antioxidative properties of diethyl blechnic, illustrating the role of ROS assays in elucidating the molecular mechanisms underlying the therapeutic effects of natural compounds (Yang S et al., 2019).

This kit is suitable for the detection of intracellular ROS by fluorescence microplate reader, flow cytometer, or a fluorescent microscope with TRITC filter.

Biochem/physiol Actions

The Fluorometric Intracellular ROS Kit provides a sensitive, one-step fluorometric assay to detect intracellular ROS (especially superoxide and hydroxyl radicals) in live cells after 1 hour incubation. ROS react with a fluorogenic sensor localized to the cytoplasm, resulting in a fluorometric product proportional to the amount of ROS present. The assay can be performed in either a 96 or 384 multiwell plate format and with detection at λex = 650/λem = 675 nm.


Still not finding the right product?

Explore all of our products under Fluorometric Intracellular Ros Kit


Storage Class

10 - Combustible liquids

WGK

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library