Skip to Content
Merck
All Photos(5)

Key Documents

M9942

Sigma-Aldrich

Anti-MAP2 Antibody

mouse monoclonal, HM-2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

Product Name

Monoclonal Anti-MAP2 antibody produced in mouse, clone HM-2, purified from hybridoma cell culture

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

HM-2, monoclonal

form

buffered aqueous solution

mol wt

antigen ~280 kDa

species reactivity

rat, chicken, human, mouse, bovine, quail

packaging

antibody small pack of 25 μL

concentration

~2 mg/mL

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
microarray: suitable
western blot: 1-2 μg/mL using rat brain extract

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAP2(4133)
mouse ... Mtap2(17756)
rat ... Map2(25595)

Looking for similar products? Visit Product Comparison Guide

General description

MAP2 is the major microtubule associated protein of brain tissue. There are three forms of MAP2; two are similarly sized with apparent molecular weights of 280 kDa (MAP2a and b) and the third with a lower molecular weight of 70 kDa (MAP2c). In the newborn rat brain, MAP2b and MAP2c are present while MAP2a is absent. Between postnatal days 10 and 20, MAP2a appears at levels equal to that of MAP2b. At the same time, the level of MAP2c drops by 10-fold. This change happens during the period when dendrite growth is completed and neurons have reached their mature morphology.
MAP2 is known to promote microtubule assembly and to form side arms on microtubules. It also interacts with neurofilaments, actin and other elements of the cytoskeleton.

Specificity

Reacts with all known forms of MAP2, namely MAP2a, MAP2b and MAP2c. Does not cross-react with other MAPs or tubulin. By immunohistochemical staining of formalin-fixed, paraffin-embedded brain tissue, the antibody shows selective labeling of dendritic trees throughout the brain.
This antibody recognizes all known forms of MAP2, namely MAP2a, MAP2b and MAP2c. It does not cross-react with other MAPs or tubulin. By immunohistochemical staining of formalin-fixed, paraffin-embedded brain tissue, the antibody shows selective labeling of dendritic trees throughout the brain.

Immunogen

rat brain microtubule-associated proteins (MAPs).

Application

Anti-MAP2 antibody has been used in immunofluorescence. It has also been used in immunohistochemistry.
May be used in various immunochemical techniques including immunoblotting, immunohistochemistry, and immunocytochemistry. Monoclonals antibodies to MAP2 are used to study the cytoskeleton structure in cells.
By immunoblotting, a working antibody concentration of 1-2 μg/ml is recommended using a rat brain extract.

Biochem/physiol Actions

MAP2 (microtubule-associated protein 2) expression is monitored to predict the disease progression in glioma. MAP2 is critical in the formation of dendritic protrusions in astrocytes and oligodendrocytes. In eukaryotes, MAP2 is essential for spindle assembly and chromosome segregation.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, and 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 C for up to one month. For extended storage, freeze in working aliquots at 20 C. Repeated freezing and thawing, or storage in frost-free freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

On and around microtubules: an overview.
Wade R H.
Molecular Biotechnology, 43(2), 177-191 (2009)
Lipolysaccharide-induced neuroinflammation is associated with Alzheimer-like amyloidogenic axonal pathology and dendritic degeneration in rats.
Deng X, et al.
Advances in Alzheimer's disease, 3(2), 78-78 (2014)
Remarkable reduction of MAP2 in the brains of scrapie-infected rodents and human prion disease possibly correlated with the increase of calpain.
Guo Y, et al.
PLoS ONE, 7, e30163-e30163 (2012)
Promotion of neuronal differentiation of neural progenitor cells by using EGFR antibody functionalized collagen scaffolds for spinal cord injury repair
Li X, et al.
Biomaterials, 34(21), 5107-5116 (2013)
Expression of EAAT2 in neurons and protoplasmic astrocytes during human cortical development
DeSilva T M, et al.
The Journal of Comparative Neurology, 520(17), 3912-3932 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service