Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.22
UNSPSC Code:
41122107
Material:
V-bottom clear wells, polystyrene
Size:
96 wells
Sterility:
sterile
Binding type:
Tissue Culture (TC)-treated surface
Feature:
lid
material
V-bottom clear wells, polystyrene
sterility
sterile
feature
lid
packaging
pack of 100
manufacturer/tradename
Greiner 651180
size
96 wells
well volume
234 μL
well working volume
40-200 μL
binding type
Tissue Culture (TC)-treated surface
General description
CELLSTAR® 96W Microplate, well working volume 40-200 μL; maximum volume 234 μL. Well surface area 0.28 cm(2).
Tissue culture treated for optimal cell attachment. Non-pyrogenic by LAL method to <0.06 EU/mL.
Application
Suitable for use with live cells. Free of detectable DNase/RNase, human DNA and pyrogens.
Other Notes
96 well microplate, with lid, solid conical (V) bottom well design, alphanumeric well coding, length: 127.76+/- 0.15 mm width: 85.48 +/- 0.15 mm curvature: approx. 200 μm.
Legal Information
CELLSTAR is a registered trademark of Greiner Bio-One GmbH
Still not finding the right product?
Explore all of our products under Greiner CELLSTAR® 96 well plates
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.