Skip to Content
Merck

Skip To

M9566

Microsomes from Liver, Pooled

from mouse(CD-1), female

Synonym(s):

extracellular matrix

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.47
UNSPSC Code:
12161501
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse (CD-1)

Quality Segment

form

liquid

packaging

vial of ~10 mg

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

mouse ... Mgst1(56615)

Biochem/physiol Actions

Liver microsomes are subcellular particles derived from the endoplasmic reticulum of hepatic cells. These microsomes are a rich source of drug metabolizing enzymes, including cytochrome P-450. Microsome pools from various sources are useful in the study of xenobiotic metabolism and drug interactions.
Source of drug metabolizing enzymes, including cytochrome P-450.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S2192S2067S2442
biological source

mouse (CD-1)

biological source

mouse (CD-1)

biological source

rat (Sprague-Dawley)

biological source

human

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100

form

liquid

form

-

form

-

form

-

packaging

vial of ~10 mg

packaging

vial of 1.0 mL

packaging

vial of ~20 mg

packaging

vial of ~20 mg

storage temp.

−70°C

storage temp.

−70°C

storage temp.

−70°C

storage temp.

−70°C

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice


Still not finding the right product?

Explore all of our products under Microsomes from Liver, Pooled


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library