Skip to Content
Merck
All Photos(1)

Documents

M9400

Sigma-Aldrich

β-Mannosidase from Helix pomatia

5-30 units/mL, ammonium sulfate suspension, crude extract

Synonym(s):

β-D-Mannoside mannohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32

form

ammonium sulfate suspension

quality

crude extract

mol wt

94 kDa

concentration

5-30 units/mL

foreign activity

β-N-acetylglucosaminidase and α-mannosidase <1.0%

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

β-Mannosidase is a lysosomal enzyme that belongs to the glycosyl hydrolase family 2.

Application

β-Mannosidase from Helix pomatia has been used:
  • in the digestion of glycans for enzymatic analysis
  • in the treatment of labeled monogalactosyl Manno-oligosaccharides
  • in non-mammalian glycosidase inhibition assays

Biochem/physiol Actions

β-Mannosidase participates in cleaving the β(1–4)-linked mannose located at the nonreducing end of N-glycosylated proteins. It is involved in the polysaccharide degradation pathway. In humans, mutations in the gene are associated with β-mannosidosis, a lysosomal storage disease. Low levels of β-Mannosidase due to mutations also lead to nystagmus, an eye disorder associated with involuntary eye movements.

Unit Definition

One unit will hydrolyze 1 μmole of p-nitrophenyl-β-D-mannopyranoside to p-nitrophenol and D-mannopyranoside per min at pH 4.0 at 25°C.

Physical form

Suspension in 3.0 M (NH4)2SO4 containing 10 mM sodium acetate, pH approx. 4.0

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pengfu Liu et al.
Applied biochemistry and biotechnology, 167(1), 73-80 (2012-04-25)
A novel biosensor strain (Escherichia coli ALM403) that responded to N-acyl homoserine lactone (AHL) was constructed using a luxR-Plux cassette as a regulatory sequence and β-mannanase as a reporter gene. Dinitrosalicylic acid method was used to detect the response of
Roneel Prakash et al.
Journal of plant physiology, 169(12), 1125-1133 (2012-06-05)
Cell walls of tomato fruit contain hemicellulosic mannans that may fulfill a structural role. Two populations were purified from cell walls of red ripe tomato tissue and named galactoglucomannan-glucuronoxylan I and II (GGM-GX I and II), respectively. Both polysaccharides not
P Wongputtisin et al.
Journal of applied microbiology, 113(4), 798-806 (2012-07-14)
To screen and select the Bacillus spp. from Tua-nao of northern Thailand for fermented corticate soybean meal (FCSBM) production. After isolation of Bacillus spp. from Tua-nao was carried out, cellulase, hemicellulases (i.e., β-mannanase and xylanase) and phytase production by isolated
A A Klyosov et al.
Carbohydrate research, 352, 65-69 (2012-03-23)
Statistical modeling was applied for describing structural features of β-(1→4)-D-galactomannans. According to the model suggested theoretical ratios of limiting degrees of locust bean, tara gum and guar gum galactomannan conversions by two β-(1→4)-mannanases of different origin (Myceliophthora thermophila and Trichoderma
Umme Afsari Zahura et al.
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology, 162(1-3), 24-33 (2012-03-01)
A β-D-mannosidase (EC 3.2.1.25) with a molecular mass of approximately 100 kDa was purified from the digestive fluid of a marine gastropod Aplysia kurodai by ammonium sulfate fractionation followed by column chromatographies on TOYOPEARL Butyl-650 M, TOYOPEARL DEAE-650 M, and

Articles

Instructions for working with enzymes supplied as ammonium sulfate suspensions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service