Skip to Content
Merck
All Photos(3)

Documents

M8765

Sigma-Aldrich

Myosmine

≥98%

Synonym(s):

3-(1-Pyrrolin-2-yl)pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10N2
CAS Number:
Molecular Weight:
146.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98%

form

solid

storage temp.

2-8°C

SMILES string

C1CN=C(C1)c2cccnc2

InChI

1S/C9H10N2/c1-3-8(7-10-5-1)9-4-2-6-11-9/h1,3,5,7H,2,4,6H2

InChI key

DPNGWXJMIILTBS-UHFFFAOYSA-N

Gene Information

Application

Tobacco alkaloid

Reactant for:
Nitrosation reactions
Peroxidation reaction with hydrogen peroxide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhao-Xiang Zhang et al.
Analytical and bioanalytical chemistry, 390(6), 1645-1652 (2008-01-30)
A novel on-column sequential preconcentration method based on the combination of field-amplified sample injection induced by acetonitrile and pseudo isotachophoresis (ITP)-acid stacking is developed for simply but efficiently concentrating alkaloid cations in a high-salt sample matrix in capillary electrophoresis. Acetonitrile
Wei Li et al.
Scientific reports, 7(1), 12126-12126 (2017-09-25)
Leaf senescence in plants is a coordinated process that involves remobilization of nutrients from senescing leaves to sink tissues. The molecular events associated with nutrient remobilization are however not well understood. In this study the tobacco system with a source-sink
Sarah Vogt et al.
Toxicology, 222(1-2), 71-79 (2006-03-01)
The incidence of esophageal adenocarcinoma is rapidly rising in Western populations. Gastroesophageal reflux disease (GERD) is thought to be one of the most important risk factors. However, the mechanisms by which GERD enhances tumor formation at the gastroesophageal junction are
Krzysztof Noworyta et al.
Analytical chemistry, 84(5), 2154-2163 (2012-03-08)
Two electropolymerizable zinc porphyrins with receptor sites tailor-designed for selective recognition of the nicotine, cotinine, or myosmine alkaloids were synthesized. These were 5-(2-phenoxyacetamide)-10,15,20-tris(triphenylamino)porphyrinato zinc(II) 1 and 5-(2,5-phenylene-bis(oxy)diacetamide)-10,15,20-tris(triphenylamino)porphyrinato zinc(II) 2 featuring one and two pendant amide side "pincers", respectively, and three
J S LaKind et al.
Risk analysis : an official publication of the Society for Risk Analysis, 19(3), 359-373 (2000-04-15)
The 16-City Study analyzed for gas-phase environmental tobacco smoke (ETS) constituents (nicotine, 3-ethenyl pyridine [3-EP], and myosmine) and for particulate-phase constituents (respirable particulate matter [RSP], ultraviolet-absorbing particulate matter [UVPM], fluorescing particulate matter [FPM], scopoletin, and solanesol). In this second of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service