Skip to Content
Merck
All Photos(1)

Documents

M8645

Sigma-Aldrich

Anti-Mouse IgG (whole molecule) antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Goat Anti-Mouse IgG, Goat IgG Antibody

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

lyophilized powder

technique(s)

Ouchterlony double diffusion: suitable

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

IgG is the main antibody type found in plasma and extracellular fluid and is expressed on B cell membrane.
Immunoglobulin G (IgG) belongs to the immunoglobulin family and is a widely expressed serum antibody. An immunoglobulin has two heavy chains and two light chains connected by a disulfide bond. It mainly helps in immune defense. It is a glycoprotein. IgG is a major class of immunoglobulin. Mouse consists of five immunoglobulin classes- IgM, IgG, IgA, IgD and IgE. Mouse IgG is further divided into five classes- IgG1, IgG2a, IgG2b and IgG3.

Immunogen

IgG purified from pooled normal mouse serum.

Application

Anti-mouse IgG (whole molecule) antibody (diluted 1:20) can be used in Indirect immunofluorescence. It can also be used as a secondary antibody in western blotting to release NC1 domain of type IV collagen. Further, it can also be used in enzyme immunoassays (50 μgram) .
Anti-mouse IgG (whole molecule) antibody (diluted 1:20) can be used in:
  • indirect immunofluorescence
  • western blotting to release NC1 domain of type IV collagen
  • enzyme immunoassays (50 μgram)

Biochem/physiol Actions

IgG regulates several functions such as phagocytosis and hence plays a pivotal role in facilitating cytological immune responses. Anti-mouse IgG (whole molecule) antibody can be used in T cell proliferation assays. It can also be used in Ouchterlony Double Diffusion and immunoelectrophoresis. Goat anti- mouse IgG (whole molecule) antibody reacts specifically with mouse IgG subclasses IgG1, IgG2a, IgG2b and IgG3. The product has also shown no reactivity to human serum proteins.
Immunoglobulin G (IgG) participates in hypersensitivity type II and type III reactions. IgG helps in opsonization, complement fixation and antibody dependent cell mediated cytotoxicity.

Other Notes

Antibody adsorbed with human serum proteins

Physical form

Lyophilized from 0.01 M sodium phosphate, 0.015 M sodium chloride, pH 7.2

Reconstitution

Reconstitute with 1 mL of 0.135 M sodium chloride.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E M Rust et al.
The Journal of clinical investigation, 103(10), 1459-1467 (1999-05-20)
The direct effects of expressing hypertrophic cardiomyopathy-associated (HCM-associated) mutant troponin T (TnT) proteins on the force generation of single adult cardiac myocytes have not been established. Replication-defective recombinant adenovirus vectors were generated for gene transfer of HCM-associated I79N and R92Q
T Keeley et al.
Zoo biology, 31(3), 275-290 (2011-03-16)
The Vancouver Island marmot (Marmota vancouverensis; VIM) is one of North America's most endangered species with fewer than 150 individuals remaining in the wild. A captive breeding program was established across four facilities in Canada as an insurance population and
Matthew Zorkau et al.
Proceedings of the National Academy of Sciences of the United States of America, 118(6) (2021-02-03)
Human mitochondria contain their own genome, mitochondrial DNA, that is expressed in the mitochondrial matrix. This genome encodes 13 vital polypeptides that are components of the multisubunit complexes that couple oxidative phosphorylation (OXPHOS). The inner mitochondrial membrane that houses these
Tim Sherlock et al.
Journal of vacuum science & technology. B, Microelectronics and nanometer structures : processing, measurement, and phenomena : an official journal of the American Vacuum Society, 29 (2011-01-01)
Corner cube retroreflectors are objects with three mutually perpendicular reflective surfaces that return light directly to its source and are therefore extremely bright and easy to detect. In this work, we have fabricated suspended corner cube retroreflectors, 5 microns in
Khalidah Ibraheem et al.
Bio-protocol, 8(13), e2907-e2907 (2018-07-05)
One fundamental property of the TNR receptor (TNFR) family relates to how 'signal quality' (the extent of receptor ligation or cross-linking) influences the outcome of receptor ligation, for instance the induction of death in tumour cells. It is unequivocal that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service