Skip to Content
Merck
All Photos(1)

Key Documents

M8558

Sigma-Aldrich

Monoclonal Anti-MDM2 antibody produced in mouse

~2 mg/mL, clone MD-219, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

MD-219, monoclonal

form

buffered aqueous solution

mol wt

antigen 90 kDa

species reactivity

mouse

should not react with

human

concentration

~2 mg/mL

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
indirect ELISA: suitable
microarray: suitable
western blot: 0.5-2 μg/mL using whole extract of transfected 293T (human embryonal kidney) cells, expressing mouse MDM2

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Mdm2(17246)

Looking for similar products? Visit Product Comparison Guide

General description

Mouse 3T3 cell double minute 2 (MDM2) gene was originally found on mouse double minute chromosomes.
Transformed mouse 3T3 cell double minute 2 (MDM2) is an E3 ligase expressed in podocytes. It is overexpressed in lymphomas.

Specificity

The antibody shows predominantly nuclear and some cytoplasmic staining in cells.

Immunogen

recombinant mouse MDM2.

Biochem/physiol Actions

In addition, to regulation MDM2 also mediates the degradation of p53. MDM2 protein has been shown to activate cell proliferation by stimulating the S-phase-inducing transcription factors, E2F1/DP1. Overexpression of MDM2 results in reduced quantities of co-expressed p53, and disruption of the p53-MDM2 interaction by mutation results in both activation and accumulation of p53. The amplification of the MDM2 gene has been demonstrated in various human neoplasms. MDM2 is frequently overexpressed in the nuclei of soft-tissue sarcomas, and to a much lesser extent in a wide variety of malignancies, such as bladder, breast, testes, and cervix cancers, and in glial cells, and a range of hematological disorders. Monoclonal antibody reacting specifically with MDM2 is an essential tool in defining the interactions and distributions of MDM2, its function in signaling pathways and in mutual regulation of p53.
Transformed mouse 3T3 cell double minute 2 (MDM2) acts as a negative regulator of the tumor suppressor p53. It also regulates the cell cycle and inflammatory activities.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dana Thomasova et al.
Journal of the American Society of Nephrology : JASN, 26(7), 1513-1523 (2014-10-29)
Murine double minute-2 (MDM2), an E3 ligase that regulates the cell cycle and inflammation, is highly expressed in podocytes. In podocyte injury, MDM2 drives podocyte loss by mitotic catastrophe, but the function of MDM2 in resting podocytes has not been
Dual Roles of MDM2 in the Regulation of p53: Ubiquitination Dependent and Ubiquitination Independent Mechanisms of MDM2 Repression of p53 Activity
Shi D, et al.
Genes & cancer, 3, 240-240 (2012)
MDM2 amplification in malignant peripheral nerve sheath tumors correlates with p53 protein expression
Wallander ML, et al.
Archives of Pathology & Laboratory Medicine, 136, 95-99 (2012)
Yun Zhang et al.
The Journal of pathology, 233(4), 380-391 (2014-05-03)
Mdm2, an E3 ubiquitin ligase, negatively regulates the tumour suppressor p53. In this study we utilized a conditional Mdm2 allele, Mdm2(FM) , and a CAG-CreER tamoxifen-inducible recombination system to examine the effects of global Mdm2 loss in adult mice. Two
P Wang et al.
Oncogene, 27(11), 1590-1598 (2007-09-11)
Mdm2, a regulator of the p53 tumor suppressor, is frequently overexpressed in lymphomas, including lymphomas that have inactivated p53. However, the biological consequences of Mdm2 overexpression in lymphocytes are not fully resolved. Here, we report that increased expression of Mdm2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service