Skip to Content
Merck

M8284

Maltose Phosphorylase from Enterococcus sp.

recombinant, expressed in E. coli, lyophilized powder

Synonym(s):

Maltose:orthophosphate 1-β-D-Glucosyltransferase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.54
EC Number:
MDL number:
Specific activity:
≥9 units/mg solid
Recombinant:
expressed in E. coli
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in E. coli

Quality Segment

form

lyophilized powder

specific activity

≥9 units/mg solid

mol wt

90 kDa by SDS-PAGE

storage temp.

−20°C

SMILES string

OC(=O)CCCCCCCCCCCCC

InChI

1S/C14H28O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14(15)16/h2-13H2,1H3,(H,15,16)

InChI key

TUNFSRHWOTWDNC-UHFFFAOYSA-N

General description

Maltose phosphorylase (MP) is a dimeric enzyme. This enzyme is classified under family 65 of the glycoside hydrolases. It is a member of the disaccharide phosphorylase family.

Application

Maltose Phosphorylase from Enterococcus sp. has been used as a component in coupled assay and ATPase activity assay to study its effects on ATPase activity, on the 90 kDa heat shock proteins (Hsp90) ATPase reaction and on background absorbance on the production of resorufin.
Maltose phosphorylase from Enterococcus has been used in a study to describe a new pathway for maltose utilization in lactic acid bacteria. It has also been used in a study to describe the transfer of glucosyl moiety of maltose to acceptors with alcoholic OH groups.

Biochem/physiol Actions

Enzymatically converts maltose to D-Glucose.
Maltose phosphorylase (MP) is a dimeric enzyme that catalyzes maltose and inorganic phosphate into β-D-glucose-1-phosphate and glucose.
Maltose phosphorylase not only transfers glucosyl moieties from maltose to other sugars but has been shown to also use phenolic compounds such as salicyl alcohol as acceptors.

Other Notes

Contains lactose.
One unit will produce 1.0 μmole of D-Glucose from maltose per minute at pH 7.0 at 30°C.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library