Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C27H38N6O9S
CAS Number:
Molecular Weight:
622.69
UNSPSC Code:
12352202
PubChem Substance ID:
MDL number:
assay
>95% (HPLC)
form
solid
storage temp.
−20°C
SMILES string
COC(=O)CCC(=O)NC(C)C(=O)NC(C)C(=O)N1CCCC1C(=O)NC(CCSC)C(=O)Nc2ccc(cc2)N(=O)=O
InChI
1S/C27H38N6O9S/c1-16(28-22(34)11-12-23(35)42-3)24(36)29-17(2)27(39)32-14-5-6-21(32)26(38)31-20(13-15-43-4)25(37)30-18-7-9-19(10-8-18)33(40)41/h7-10,16-17,20-21H,5-6,11-15H2,1-4H3,(H,28,34)(H,29,36)(H,30,37)(H,31,38)
InChI key
LUUQUDLEABXXQL-UHFFFAOYSA-N
Gene Information
human ... CTSG(1511)
mouse ... CTSG(13035)
rat ... CTSG(290257)
General description
Colorimetic substrate for cathepsin G.
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.