Skip to Content
Merck

M6949

Madecassoside from Centella asiatica

≥95% (HPLC)

Synonym(s):

Madecassoside, Asiaticoside A, Redermic

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C48H78O20
CAS Number:
Molecular Weight:
975.12
UNSPSC Code:
12352212
EC Number:
252-076-1
NACRES:
NA.25
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder

color

white to off-white

solubility

H2O: 1 mg/mL, clear, colorless

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

O1C(C(C(C(C1C)O)O)O)OC2C(OC(C(C2O)O)OCC3OC(C(C(C3O)O)O)OC(=O)C54C(C(C(CC5)C)C)C6=CCC7C8(C(C(CC7(C6(CC4)C)C)O)C(C(C(C8)O)O)(CO)C)C)CO

InChI

1S/C48H78O20/c1-19-10-11-48(13-12-46(6)22(28(48)20(19)2)8-9-27-44(4)14-24(52)39(61)45(5,18-50)38(44)23(51)15-47(27,46)7)43(62)68-42-35(59)32(56)30(54)26(66-42)17-63-40-36(60)33(57)37(25(16-49)65-40)67-41-34(58)31(55)29(53)21(3)64-41/h8,19-21,23-42,49-61H,9-18H2,1-7H3

InChI key

BNMGUJRJUUDLHW-UHFFFAOYSA-N

General description

Madecassoside is one of the major pentacyclic triterpenoid saponin compounds extracted from Centella asiatica.

Application

Madecassoside has been used to study its cytotoxic effects on mouse macrophage cell lines (J774A.1). It has also been used as a potential inhibitor of serine racemase (SR) for in vitro and ex vivo studies.

Biochem/physiol Actions

Madecassoside has a wide range of reported biological activities including, anti-inflammatory, wound healing, and antioxidant activities. It has been reported to suppress LPS-induced TNF-alpha production via inhibition of ERK, p38, and NF-kappaB activity.
Madecassoside has skin hydration properties. It is a well-known collagen inducer that exerts anti-aging and anti-rheumatoid arthritis effects. Madecassoside also elicits neuroprotective activities.
Madecassoside is a triterpenoid compound found in Centella asiatica (Gotu kola), a medicinal herb used in traditional Chinese medicine (TCM), Ayurvedic medicine, and traditional African medicine. Madecassoside has a wide range of reported biological activities including, anti-inflammatory, wound healing, and anti-oxidant activities. It has been reported to suppress LPS-induced TNF-alpha production via inhibition of ERK, p38, and NF-kappaB activity.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Marek Haftek et al.
Experimental dermatology, 17(11), 946-952 (2008-05-28)
Skin ageing is a complex process determined by the genetic endowment of individual and environmental factors, such as sun exposure. The effects of skin ageing are mostly encountered in the superficial dermis and in the epidermis. We have previously demonstrated
Gui-Gui Li et al.
Yao xue xue bao = Acta pharmaceutica Sinica, 42(5), 475-480 (2007-08-21)
This study is to investigate if madecassoside can protect against myocardial reperfusion injury in rabbit heart in vivo. The ischemia reperfusion model was established. Left ventricular function and ECG were monitored at the ischemia and reperfusion period. The infarct areas
Mingzhe Gao et al.
Se pu = Chinese journal of chromatography, 26(3), 362-365 (2008-08-30)
A high performance liquid chromatographic (HPLC) method was successfully used to separate asiaticoside and madecassoside from the extract of Centella asiatica (L.) Urb. The raw material containing 20% asiaticoside and 45% madecassoside was dissolved in methanol and used for the



Global Trade Item Number

SKUGTIN
M6949-50MG04061834061465
M6949-10MG04061834061458