Skip to Content
Merck
All Photos(1)

Documents

M6690

Sigma-Aldrich

MDL 28170

≥90% (TLC)

Synonym(s):

Calpain Inhibitor III

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H26N2O4
CAS Number:
Molecular Weight:
382.45
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥90% (TLC)

composition

Peptide Content, >90%

solubility

DMSO: 26 mg/mL
H2O: insoluble
methanol: soluble

shipped in

wet ice

storage temp.

2-8°C

SMILES string

[H]C(=O)C(Cc1ccccc1)NC(=O)[C@@H](NC(=O)OCc2ccccc2)C(C)C

InChI

1S/C22H26N2O4/c1-16(2)20(24-22(27)28-15-18-11-7-4-8-12-18)21(26)23-19(14-25)13-17-9-5-3-6-10-17/h3-12,14,16,19-20H,13,15H2,1-2H3,(H,23,26)(H,24,27)/t19?,20-/m0/s1

InChI key

NGBKFLTYGSREKK-ANYOKISRSA-N

Gene Information

Amino Acid Sequence

Z-Val-Phe-al

Application

MDL 28170 has been used as a calpain inhibitor:
  • to study its effects on neuroinflammation and necroptosis in rat model of cardiac arrest
  • to analyze its effects on the degradation of neuronal nitric oxide synthase (nNOS) in beef semimembranosus muscle
  • to study its protective effects against traumatic brain injury and survival of bone marrow-derived mesenchymal stem cells (BMSCs) in rat brain

Biochem/physiol Actions

MDL 28170 exhibits neuroprotective effects against spinal cord injury, focal cerebral ischemia, and neonatal hypoxia-ischemia in rats. It also shows anti-inflammatory and anti-neurodegenerative properties. MDL 28170 crosses the blood-brain barrier and penetrates readily through the cell membranes.
MDL 28170 is a potent cell permeable calpain I and II inhibitor; reduces capsaicin-mediated cell death in cultured dorsal root ganglion neurons. Reduced occurrence of apoptosis in H2O2 and A23187 treated PC12 cells. γ-secretase-inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wen-Yan Wang et al.
International immunopharmacology, 93, 107377-107377 (2021-02-01)
Cerebral ischemia-reperfusion injury (CIRI) is the leading cause of poor neurological prognosis after cardiopulmonary resuscitation (CPR). We previously reported that the extracellular signal-regulated kinase (ERK) activation mediates CIRI. Here, we explored the potential ERK/calpain-2 pathway role in CIRI using a
Jiangnan Hu et al.
Stem cell research & therapy, 10(1), 96-96 (2019-03-17)
Studies have shown that transplantation of bone marrow-derived mesenchymal stem cells (BMSCs) protects against brain damage. However, the low survival number of transplanted BMSCs remains a pertinent challenge and can be attributed to the unfavorable microenvironment of the injured brain.
Stephanie N Thompson et al.
Journal of neurotrauma, 27(12), 2233-2243 (2010-09-30)
The cytoskeletal and neuronal protective effects of early treatment with the blood-brain barrier- and cell-permeable calpain inhibitor MDL-28170 was examined in the controlled cortical impact (CCI) traumatic brain injury (TBI) model in male CF-1 mice. This was preceded by a
Lorelei B Silverman-Gavrila et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(5), 1975-1990 (2013-02-01)
Low-frequency depression (LFD) of transmitter release occurs at phasic synapses with stimulation at 0.2 Hz in both isolated crayfish (Procambarus clarkii) neuromuscular junction (NMJ) preparations and in intact animals. LFD is regulated by presynaptic activity of the Ca(2+)-dependent phosphatase calcineurin
Amy L Clark et al.
Scientific reports, 7(1), 5611-5611 (2017-07-19)
Pro-inflammatory cytokines are important mediators of islet inflammation, leading to beta cell death in type 1 diabetes. Although alterations in both endoplasmic reticulum (ER) and cytosolic free calcium levels are known to play a role in cytokine-mediated beta cell death

Articles

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service