Skip to Content

M6686

Greiner 384 well plates, polystyrene

medium-binding

, flat bottom clear polystyrene wells

Synonym(s):

384 multiwell plates, 384 well microplates, 384 well microtiter plates, 384 well plates, Greiner 781095

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.22
UNSPSC Code:
41122107
Material:
flat bottom clear polystyrene wells, white polystyrene
Size:
384 wells
Sterility:
non-sterile
Binding type:
medium binding surface
Feature:
lid: no
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

flat bottom clear polystyrene wells, white polystyrene

sterility

non-sterile

feature

lid: no

packaging

case of 40

manufacturer/tradename

Greiner 781095

maximum volume

131 μL

size

384 wells

working volume

110 μL

binding type

medium binding surface

General description

384 flat bottom, medium binding, microplate,with an optimal working volume of 15-110 μL; maximum well capacity of 131 μL. Growth area: 10 mm(2)
Greiner multiwell plates are available in the standard multiwell plate footprint and offer excellent optical properties. These plates are DNAse /RNAse free and non-pyrogenic as deteremined by LAL method (< 0.06 EU/mL). Appropriate for general purposes, such as optical measurements, photography, and microscopy, due to the high transparency of polystyrene plates.

Application

White plates minimize cross talk in luminescence procedures. μClear film improves Luminescence; Fluorescence; Cell monitoring; Transmission; and Imaging by reducing background, cross-talk and autofluorescence. Free of detectable DNase/RNase, human DNA and pyrogens.

Other Notes

384 flat well microtiter plate, without lid, rounded square well design, alphanumeric well coding; length: 127.76 +/- 0.15 mm; width: 85.48 +/- 0.15 mm; curvature: approx.150 μm.

Legal Information



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library