Skip to Content
Merck
All Photos(2)

Key Documents

M6635

Sigma-Aldrich

N-(2-Mercaptopropionyl)glycine

Synonym(s):

Tiopronin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(SH)CONHCH2COOH
CAS Number:
Molecular Weight:
163.19
Beilstein:
1859822
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

Assay

≥98% (TLC)

form

powder

color

white

mp

98-100 °C

application(s)

cell analysis
peptide synthesis

SMILES string

CC(S)C(=O)NCC(O)=O

InChI

1S/C5H9NO3S/c1-3(10)5(9)6-2-4(7)8/h3,10H,2H2,1H3,(H,6,9)(H,7,8)

InChI key

YTGJWQPHMWSCST-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-(2-Mercaptopropionyl)glycine or Tiopronin has been used:
  • in the preparation of polyethylenimine (PEI)-coated gold microparticles (PEI-gold) for biolistic delivery of nucleic acids
  • to study the role of reactive oxygen species (ROS) at the reperfusion stage in in vivo isoflurane preconditioning-induced neuroprotection
  • as a ROS inhibitor to study its effect on lysophosphatidylcholine (LPC)-induced inflammasome activation

Biochem/physiol Actions

N-(2-Mercaptopropionyl)glycine (NMPG) is a thiol compound associated with autoimmune hypoglycemia. It is a diffusible antioxidant and reduces the severity of colonic injury. NMPG also relieves myeloperoxidase activity and stimulates hypoxia-inducible factor-1 (HIF-1) - vascular endothelial growth factor (VEGF) pathway for ulcer healing.
N-(2-Mercaptopropionyl)glycine is a free radical scavenger.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soohwan Yum et al.
Biochemical and biophysical research communications, 443(3), 1008-1013 (2013-12-24)
We investigated anti-colitic effects of N-(2-mercaptopropionyl)-glycine (NMPG), a diffusible antioxidant, in TNBS-induced rat colitis model and a potential molecular mechanism underlying the pharmacologic effect of the antioxidant. NMPG alleviated colonic injury and effectively lowered myeloperoxidase activity. Moreover, NMPG substantially attenuated
Pyritinol
Meyler's Side Effects of Drugs: The International Encyclopedia of Adverse Drug Reactions and Interactions, 2988-2989 (2006)
Sang Hyun Lee et al.
Brain research, 1723, 146405-146405 (2019-08-28)
In this in vivo and in vitro study, we aimed to investigate whether isoflurane preconditioning-induced neuronal protection is mediated by reactive oxygen species (ROS) signaling at the reperfusion stage. In the in vivo study, Sprague-Dawley rats were subjected to middle
Wei Chen et al.
Journal of cardiovascular pharmacology, 73(5), 265-271 (2019-05-15)
Emulsified isoflurane (EI) has been shown to alleviate myocardial ischemia-reperfusion (IR) injury. However, previous reports have not been focused on the underlying mechanism. We used models of IR injury in Langendorff-isolated rat hearts to determine the relationship between the mechanism
Christoph Becker-Pauly et al.
Molecular & cellular proteomics : MCP, 10(9), M111-M111 (2011-06-23)
Astacins are secreted and membrane-bound metalloproteases with clear associations to many important pathological and physiological processes. Yet with only a few substrates described their biological roles are enigmatic. Moreover, the lack of knowledge of astacin cleavage site specificities hampers assay

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service