Skip to Content
Merck
All Photos(2)

Key Documents

M4910

Sigma-Aldrich

1-Methylhistamine dihydrochloride

≥98% (TLC), powder

Synonym(s):

1-Methyl-4-(β-aminoethyl)imidazole dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11N3 · 2HCl
CAS Number:
Molecular Weight:
198.09
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Product Name

1-Methylhistamine dihydrochloride, ≥98% (TLC), powder

Quality Level

Assay

≥98% (TLC)

form

powder

solubility

water: 49.00-51.00 mg/mL, clear, colorless to faintly yellow

storage temp.

2-8°C

SMILES string

Cl[H].Cl[H].Cn1cnc(CCN)c1

InChI

1S/C6H11N3.2ClH/c1-9-4-6(2-3-7)8-5-9;;/h4-5H,2-3,7H2,1H3;2*1H

InChI key

AGXVEALMQHTMSW-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

1-Methylhistamine dihydrochloride has been used as a standard in the evaluation of histamine levels in aqueous humor of glaucoma patients by liquid chromatography. It has also been used as a substrate to determine amine oxidase activity.

Biochem/physiol Actions

1-Methylhistamine is present in the brain and is a metabolite of histamine that is metabolized by histamine N-methyltransferase. It acts as a biomarker for histaminergic system activity in the cerebrospinal fluid of patients suffering from Alzheimer′s disease (AD), hypersomnia, and other neurological conditions.
Major metabolite of histamine by histamine N-methyltransferase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Hashimoto et al.
Acta anaesthesiologica Scandinavica, 42(7), 858-863 (1998-08-12)
Histamine is most densely distributed in the hypothalamus and has an important effect on consciousness or wakefulness. It has been little considered whether general anesthetics could exert their effects on hypothalamic histamine metabolism. The present study was conducted to investigate
M Motawaj et al.
The Journal of pharmacology and experimental therapeutics, 336(2), 479-487 (2010-11-09)
We previously reported that some N-methyl-D-aspartate (NMDA)-receptor antagonists enhanced histamine neuron activity in rodents. Here, we have investigated the effects of memantine, an NMDA-receptor antagonist used for the treatment of Alzheimer's disease, on histaminergic neurotransmission. In vitro, memantine antagonized native
Yasuhisa Miyamoto et al.
Analytical biochemistry, 334(1), 89-96 (2004-10-07)
An improved high-performance liquid chromatography (HPLC) method was developed for simultaneous analysis of histamine (HA) and tele-methylhistamine (tele-MHA) levels in mouse and rat brain. The method consists of a solid-phase extraction (SPE) and subsequent HPLC with postcolumn derivatization of the
Inability to replicate cerebrospinal fluid histamine deficits in the primary hypersomnias: a back to the drawing board moment.
David B Rye
Sleep, 35(10), 1315-1317 (2012-10-02)
A Burban et al.
The Journal of pharmacology and experimental therapeutics, 332(3), 912-921 (2009-12-17)
Histamine potentiates activation of native and recombinant N-methyl-d-aspartate receptors (NMDARs), but its mechanisms of action and physiological functions in the brain remain controversial. Using four different models, we have further investigated the histamine-induced potentiation of various NMDAR-mediated responses. In single

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service