Skip to Content
Merck

M4565

Matrix Metalloproteinase-7 human

~0.1 mg/mL, recombinant, expressed in E. coli, buffered aqueous solution

Synonym(s):

MMP-7, Matrilysin-1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32
EC Number:
MDL number:
Specific activity:
5,000 units/mg protein
Recombinant:
expressed in E. coli
Concentration:
~0.1 mg/mL
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in E. coli

Quality Segment

form

buffered aqueous solution

specific activity

5,000 units/mg protein

mol wt

20 kDa

concentration

~0.1 mg/mL

UniProt accession no.

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MMP7(4316)

General description

Matrix metalloproteinase-7 (MMP7) also referred to as matrilysin is encoded by the gene mapped to human chromosome 11q21-q22. MMP7 is a smallest member of MMP enzyme family and is highly expressed in cancer cells. MMP7 is exclusively released by epithelial cells.

Application

Matrix metalloproteinase-7 (MMP7) human has been used in in silico analysis of MMP-7 proteolysis of perlecan.

Biochem/physiol Actions

Matrix metalloproteinase-7 (MMP-7) catalyzes the cleavage of extracellular matrix (ECM) proteins such as proteoglycans, fibronectin, entactin, laminin, collagen III/ IV/ V/ IX/ X/ XI, type I/ II/ IV/ V gelatins, and elastin. Matrix metalloproteinase-7 facilitates initial stages of tumor progression. Activated MMP7 plays a vital role in human colorectal cancer (CRC) liver metastases. Overexpression of MMP7 is observed in various human cancers, such as breast, lung, prostate, esophagus, stomach, endometrium, and ovarian carcinomas, as well as esophageal squamous cell carcinomas. Increased expression of MMP7 is also associated with pathogenesis of demyelinating multiple sclerosis (MS) lesions. Both in silico and in vitro studies show that MMP7 degrades glycosylated and basement membrane bound perlecan and plays a vital role in prostate cancer cell invasion.

Physical form

Solution in 10 mM HEPES, 5 mM CaCl2, 150 mM NaCl

Analysis Note

One unit of enzyme activity is defined as the digestion of 1 μg of azocoll/min. at pH 7.5 and 37 °C.


flash_point_f

Not applicable

flash_point_c

Not applicable

wgk

WGK 1

Storage Class

12 - Non Combustible Liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library