Skip to Content
Merck

M4255

3-Methoxy-L-tyrosine monohydrate

≥98% (HPLC)

Synonym(s):

4-Hydroxy-3-methoxy-L-phenylalanine, L-3-O-Methyl-DOPA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C10H13NO4 H2O
CAS Number:
Molecular Weight:
229.23
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

3-Methoxy-L-tyrosine monohydrate, powder

assay

≥98% (HPLC)

Quality Segment

form

powder

application(s)

detection

storage temp.

2-8°C

SMILES string

COc1cc(C[C@H](N)C(O)=O)ccc1O

InChI

1S/C10H13NO4/c1-15-9-5-6(2-3-8(9)12)4-7(11)10(13)14/h2-3,5,7,12H,4,11H2,1H3,(H,13,14)/t7-/m0/s1

InChI key

PFDUUKDQEHURQC-ZETCQYMHSA-N

Biochem/physiol Actions

3-Methoxy-L-tyrosine is a metabolite of L-Dopa.
The major metabolite of L-DOPA via catechol O-methyltransferase (COMT). 3-methoxy-L-tyrosine is neither a substrate for nor an inhibitor of L-amino acid decarboxylase activity. Inhibition of COMT enhances the anti-Parkinson activity of L-DOPA.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library