Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352204
NACRES:
NA.28
Product Name
Mutanolysin Assay Substrate, (Lyophilized suspension of washed Enterococcus faecalis STF-3 (ATCC 12984) cell walls.)
Quality Segment
form
(Lyophilized suspension of washed Enterococcus faecalis STF-3 (ATCC 12984) cell walls.)
packaging
vial of 10 assays
shipped in
wet ice
storage temp.
−20°C
Application
The activity of mutanolysin can be determined by hydrolysis of the rehydrated cell suspension in a continuous spectrophotometric assay.
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Datasheet
Global Trade Item Number
| SKU | GTIN |
|---|---|
| M3440-1VL | 04061838164025 |