Skip to Content
Merck

M3003

Myokinase from rabbit muscle

ammonium sulfate suspension, 1,500-3,000 units/mg protein (biuret)

Synonym(s):

ATP: AMP phosphotransferase, Adenylate Kinase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352204
eCl@ss:
32160410
EC Number:
232-743-3
NACRES:
NA.54
MDL number:
EC Number:
Specific activity:
1,500-3,000 units/mg protein (biuret)
Biological source:
rabbit muscle
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit muscle

Quality Level

form

ammonium sulfate suspension

specific activity

1,500-3,000 units/mg protein (biuret)

mol wt

21 kDa

foreign activity

3-phosphoglyceric phosphokinase ≤0.05%, lactic dehydrogenase and pyruvate kinase ≤0.01%

storage temp.

2-8°C

General description

Research area: Cell Signaling
Myokinase, also called Adenylate kinase or ADK, is a phosphotransferase enzyme. It belongs to the family of nucleoside monophosphate kinases. Myokinase is an enzyme that can be found in skeletal muscle and acts as a phosphotransferase agitator.

Application

Myokinase has been used:

  • in a study to assess its reaction with the mercurials p-hydroxymercuribenzoate and p-mercuribenzenesulfonate
  • in a study to investigate the ligand-binding properties of its peptide fragments
  • as one of the components in the ATP generation solution for the quantification of 3’3’-c-di-AMP using an AMP-Luciferase assay

Biochem/physiol Actions

Myokinase or adenylate kinases, play a crucial role in regulating energy metabolism within cells and balancing adenine nucleotide ratios in different cellular compartments.

Myokinase is a key enzyme involved with adenine nucleotide metabolism. The action of myokinase insures that the adenine nucleotides (ATP, ADP, and AMP) are maintained in equilibrium in the cytosol.
Myokinase catalyzes the following reaction:ADP + ADP → ATP + AMP
Rabbit muscle myokinase can be inactivated by treatment with dithiobis-(2-nitrobenzoic) acid which alters methionine residues critical to myokinase activity.

Physical form

Suspension in 3.2 M (NH4)2SO4 solution containing 0.001 M EDTA, pH 6.0

Other Notes

One unit will convert 2.0 μmoles of ADP to ATP+AMP per min at pH 7.6 at 37°C.


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


Myokinase, ATP-AMP Transphosphorylase.
Noda, L., and Kuby, S.A.
Methods in Enzymology, 6, 223-230 (1962)
The sulfhydryl groups of rabbit muscle adenosine triphosphate-adenosine monophosphate phosphotransferase. Activity of enzyme treated with mercurials.
L F Kress et al.
The Journal of biological chemistry, 241(10), 2293-2300 (1966-05-25)
Likui Feng et al.
Nature communications, 12(1), 4912-4912 (2021-08-15)
Polyketide synthase (PKS) and nonribosomal peptide synthetase (NRPS) hybrid systems typically use complex protein-protein interactions to facilitate direct transfer of intermediates between these multimodular megaenzymes. In the canal-associated neurons (CANs) of Caenorhabditis elegans, PKS-1 and NRPS-1 produce the nemamides, the only known



Global Trade Item Number

SKUGTIN
M3003-1KU04061832924526
M3003-2.5KU04061834046295
M3003-10KU04061832924519
M3003-5KU04061832924540