Skip to Content
Merck
All Photos(1)

Key Documents

M2699

Sigma-Aldrich

Marimastat

≥98% (HPLC), solid, MMP inhibitor

Synonym(s):

BB2516, (2S,3R)-N4-[(1S)-2,2-Dimethyl-1-[(methylamino)carbonyl] propyl]-N1,2-dihydroxy-3-(2-methylpropyl)butanediamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H29N3O5
CAS Number:
Molecular Weight:
331.41
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Marimastat, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

solid

solubility

DMSO: ≥20 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

CNC(=O)[C@@H](NC(=O)[C@H](CC(C)C)[C@H](O)C(=O)NO)C(C)(C)C

InChI

1S/C15H29N3O5/c1-8(2)7-9(10(19)13(21)18-23)12(20)17-11(14(22)16-6)15(3,4)5/h8-11,19,23H,7H2,1-6H3,(H,16,22)(H,17,20)(H,18,21)/t9-,10+,11-/m1/s1

InChI key

OCSMOTCMPXTDND-OUAUKWLOSA-N

Gene Information

Related Categories

Application

Marimastat has been used as an inhibitor of:
  • metalloproteinase 2/9 (MMP2/9), to study its effects on exercise-mediated interleukin-6 (IL-6) release in mice[1]
  • metalloproteinase, to determine protease activity in Pseudomonas aeruginosa cultures[2]
  • metalloproteinase 10 (MMP10), to study its effect on monoclonal antibody H3 binding to MMP10[3]

Biochem/physiol Actions

Marimastat is a broad spectrum matrix metalloprotease (MMP) inhibitor

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Ohshima et al.
Journal of dental research, 89(11), 1315-1321 (2010-08-27)
The underlying mechanism and the therapeutic regimen for the transition of reversible gingivitis to irreversible periodontitis are unclear. Since transforming growth factor (TGF)-β has been implicated in differentially regulated gene expression in gingival fibroblasts, we hypothesized that TGF-β signaling is
Angela Sandri et al.
Virulence, 9(1), 1008-1018 (2018-06-26)
Cystic fibrosis (CF) lung infection is a complex condition where opportunistic pathogens and defective immune system cooperate in developing a constant cycle of infection and inflammation. The major pathogen, Pseudomonas aeruginosa, secretes a multitude of virulence factors involved in host
Vincent E de Meijer et al.
PloS one, 5(6), e11256-e11256 (2010-07-02)
Liver fibrosis is characterized by excessive synthesis of extracellular matrix proteins, which prevails over their enzymatic degradation, primarily by matrix metalloproteinases (MMPs). The effect of pharmacological MMP inhibition on fibrogenesis, however, is largely unexplored. Inflammation is considered a prerequisite and
Timothy W Failes et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 13(10), 2974-2982 (2006-12-16)
We report a potential means of selectively delivering matrix metalloproteinase (MMP) inhibitors to target tumour sites by use of a bioreductively activated Co(III) carrier system. The carrier, comprising a Co(III) complex of the tripodal ligand tris(methylpyridyl)amine (tpa), was investigated with
Victor A Levin et al.
Journal of neuro-oncology, 78(3), 295-302 (2006-04-26)
Because raised matrix metalloprotease (MMP) levels are associated with glioma invasion and angiogenesis, we tested the efficacy of marimastat (MT) an orally active drug that can reduce MMP levels, in patients with gliomas. A total of 162 patients with intracranial

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service