Skip to Content
Merck
All Photos(1)

Key Documents

M2696

Sigma-Aldrich

Metyrapone

≥98% (HPLC), solid

Synonym(s):

2-Methyl-1,2-di-3-pyridyl-1-propanone, Metyrapone, Su-4885

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14N2O
CAS Number:
Molecular Weight:
226.27
Beilstein:
163023
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

sterility

non-sterile

Quality Level

Assay

≥98% (HPLC)

form

solid

technique(s)

toxicology assay: suitable

mp

52-55 °C (lit.)

solubility

H2O: <2 mg/mL
DMSO: >20 mg/mL

originator

Novartis

shipped in

ambient

storage temp.

room temp

SMILES string

CC(C)(c1cccnc1)C(=O)c2cccnc2

InChI

1S/C14H14N2O/c1-14(2,12-6-4-8-16-10-12)13(17)11-5-3-7-15-9-11/h3-10H,1-2H3

InChI key

FJLBFSROUSIWMA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Metyrapone is a glucocorticoid synthesis inhibitor; a standard in assessing adrenal gland function especially in response to stress, as well as pituitary gland function. In addition to acting as a glucocorticoid synthesis inhibitor, Metyrapone also inhibits cytochrome P450-mediated prostaglandin ω/ω-1 hydroxylase activity and impairs learning and memory. Data shows results in activation of the sleep EEG and a robust decrease in quantitative delta sleep.

Features and Benefits

This compound was developed by Novartis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Irit Akirav et al.
Learning & memory (Cold Spring Harbor, N.Y.), 11(2), 188-195 (2004-04-01)
Emotionally charged experiences alter memory storage via the activation of hormonal systems. Previously, we have shown that compared with rats trained for a massed spatial learning task in the water maze in warm water (25 degrees C), animals that were
Angus Lindsay et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 35(4), e21489-e21489 (2021-03-19)
Psychosocial stressors can cause physical inactivity, cardiac damage, and hypotension-induced death in the mdx mouse model of Duchenne muscular dystrophy (DMD). Because repeated exposure to mild stress can lead to habituation in wild-type mice, we investigated the response of mdx
Therese Koal et al.
The Journal of steroid biochemistry and molecular biology, 129(3-5), 129-138 (2012-01-03)
In order to overcome many limitations of immunoassays, high performance liquid chromatography-tandem mass spectrometry (HPLC-MS/MS) has the potential to find its place in the clinical laboratory medicine for quantification of steroid hormones. A prerequisite for the application of a new
Sonja C Schriever et al.
The Journal of clinical investigation, 130(11), 6093-6108 (2020-08-12)
Recent genome-wide association studies (GWAS) identified DUSP8, encoding a dual-specificity phosphatase targeting mitogen-activated protein kinases, as a type 2 diabetes (T2D) risk gene. Here, we reveal that Dusp8 is a gatekeeper in the hypothalamic control of glucose homeostasis in mice
Xavier Belda et al.
Hormones and behavior, 62(4), 515-524 (2012-09-19)
A single exposure to some severe stressors causes sensitization of the hypothalamic-pituitary-adrenal (HPA) response to novel stressors. However, the putative factors involved in stress-induced sensitization are not known. In the present work we studied in adult male rats the possible

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service