Skip to Content
Merck

M227

MRS 1191

solid

Synonym(s):

3-Ethyl-5-benzyl-2-methyl-4-phenylethynyl-6-phenyl-1,4-(±)-dihydropyridine-3,5-dicarboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C31H27NO4
CAS Number:
Molecular Weight:
477.55
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Form:
solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solid

Quality Segment

color

white

solubility

DMSO: >10 mg/mL, ethanol: >10 mg/mL, H2O: insoluble

storage temp.

−20°C

SMILES string

CCOC(=O)C1=C(C)NC(c2ccccc2)=C(C1C#Cc3ccccc3)C(=O)OCc4ccccc4

InChI

1S/C31H27NO4/c1-3-35-30(33)27-22(2)32-29(25-17-11-6-12-18-25)28(26(27)20-19-23-13-7-4-8-14-23)31(34)36-21-24-15-9-5-10-16-24/h4-18,26,32H,3,21H2,1-2H3

InChI key

SNVFDPHQAOXWJZ-UHFFFAOYSA-N

Biochem/physiol Actions

MRS 1191 is putative A3 adenosine receptor antagonist, highly selective for human A3 receptor vs human A1 receptor. MRS 1067, MRS 1191 and MRS 1220 were found to be competitive in saturation binding studies using the agonist radioligand [125I]AB-MECA at cloned human brain A3 receptors expressed in HEK-293 cells. Antagonism was demonstrated in functional assays consisting of agonist-induced inhibition of adenylate cyclase and the stimulation of binding of [35S]guanosine 5′-O-(3-thiotriphosphate) ([35S]GTP-gamma-S) to the associated G-proteins. Activation of the human A3 receptor in A3R-CHO results in markedly impaired cell cycle progression, suggesting an important role for this adenosine receptor subtype in cell cycle regulation and cell growth. Activation of adenosine A3 receptors by Cl-IBMECA (100 nM) increased the magnitude of theta-burst induced LTP (from 1.2+/-0.6% in the control solution to 25.5+/-0.8% in the presence of Cl-IBMECA) and attenuated LTD (from 30.0+/-5.5% decrease in the control solution to 13.6+/-6.6% decrease in the presence of Cl-IBMECA). The selective adenosine A3 receptor antagonist, MRS 1191 (5-10 μM), prevented the effects of Cl-IBMECA. These findings indicate a functional role for adenosine A3 receptors in the modulation of synaptic plasticity.
MRS 1191 is putative A3 adenosine receptor antagonist, highly selective for human A3 receptor vs human A1 receptor.

Disclaimer

Photosensitive


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library