Skip to Content
Merck

M2265

1-Octanoyl-rac-glycerol

≥99%

Synonym(s):

Glyceryl 1-monooctanoate, 1-Capryloyl-rac-glycerol, Monocaprylin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C11H22O4
CAS Number:
Molecular Weight:
218.29
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352211
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥99%

form

powder

functional group

ester

lipid type

neutral glycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCC(=O)OCC(O)CO

InChI

1S/C11H22O4/c1-2-3-4-5-6-7-11(14)15-9-10(13)8-12/h10,12-13H,2-9H2,1H3

InChI key

GHBFNMLVSPCDGN-UHFFFAOYSA-N

Application


  • Molecular Simulations and Markov State Modeling of Quorum Sensing Regulators: Research revealed the inactive form of the quorum sensing regulator SdiA in Escherichia coli, offering insights into bacterial communication mechanisms that could be influenced by lipid-based signaling molecules such as 1-Octanoyl-rac-glycerol (Grabski et al., 2021).

Biochem/physiol Actions

Monocaprylin (1-octanoyl-rac-glycerol) is used as an antimicrobial monoglycerin with potential to treat bacteria D congolensis and campylobacter infections and to inactivate E. coli, Listeria and Salmonella on biosurfaces.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library