Skip to Content
Merck
All Photos(1)

Key Documents

M1777

Sigma-Aldrich

N-Methyl-1-deoxynojirimycin

≥98%

Synonym(s):

1,5-Dideoxy-1,5-imino-1-methyl-D-sorbitol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H15NO4
CAS Number:
Molecular Weight:
177.20
Beilstein:
1524564
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.32

biological source

synthetic (organic)

Quality Level

Assay

≥98%

form

powder

solubility

methanol: 10 mg/mL, clear, colorless

antibiotic activity spectrum

viruses

Mode of action

enzyme | inhibits

storage temp.

−20°C

SMILES string

CN1C[C@H](O)[C@@H](O)[C@H](O)[C@H]1CO

InChI

1S/C7H15NO4/c1-8-2-5(10)7(12)6(11)4(8)3-9/h4-7,9-12H,2-3H2,1H3/t4-,5+,6-,7-/m1/s1

InChI key

AAKDPDFZMNYDLR-XZBKPIIZSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: glucosamine

Biochem/physiol Actions

Interferes with metabolism of N-linked glycoproteins by inhibition of glucosidase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diego A Caraballo et al.
G3 (Bethesda, Md.), 10(2), 755-768 (2019-12-05)
UDP- glucose: glycoprotein glucosyltransferase (UGGT) is a protein that operates as the gatekeeper for the endoplasmic reticulum (ER) quality control mechanism of glycoprotein folding. It is known that vertebrates and Caenorhabditis genomes harbor two uggt gene copies that exhibit differences
N Y Marcus et al.
The Journal of biological chemistry, 275(3), 1987-1992 (2000-01-15)
It is now well known that the addition and trimming of oligosaccharide side chains during post-translational modification play an important role in determining the fate of secretory, membrane, and lysosomal glycoproteins. Recent studies have suggested that trimming of oligosaccharide side
K Murakami et al.
Journal of bacteriology, 176(9), 2635-2639 (1994-05-01)
A Mucor pusillus mutant defective in asparagine-linked glycosylation was found in our stock cultures. This mutant, designated 1116, secreted aspartic proteinase (MPP) in a less-glycosylated form than that secreted by the wild-type strain. Analysis of enzyme susceptibility, lectin binding, and
M Arai et al.
British journal of pharmacology, 133(5), 635-642 (2001-06-29)
Preischaemic treatment with N-methyl-1-deoxynojirimycin (MOR-14), an alpha-1,6-glucosidase inhibitor, attenuates glycogenolysis and lactate accumulation during ischaemia and markedly reduces infarct size in rabbit hearts. In the present study, we have investigated whether protein kinase C (PKC), a principal mediator of ischaemic
G C Trudel et al.
Biochemical and biophysical research communications, 184(1), 125-130 (1992-04-15)
The glycoprotein processing inhibitors bromoconduritol and N-methyl-1-deoxynojirimycin inhibit myoblast fusion and differentiation, suggesting the critical involvement of one or more glycoproteins in the control of skeletal myogenesis. In the present study we have examined the effect of inhibitors of glycoprotein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service