Skip to Content
Merck
All Photos(2)

Documents

M1651

Sigma-Aldrich

Sodium molybdate dihydrate

≥99.5%, suitable for plant cell culture

Synonym(s):

Molybdic acid sodium salt dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2MoO4 · 2H2O
CAS Number:
Molecular Weight:
241.95
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.71

Assay

≥99.5%

reaction suitability

core: molybdenum

technique(s)

cell culture | plant: suitable

mp

100 °C (dec.) (lit.)

SMILES string

O.O.[Na+].[Na+].[O-][Mo]([O-])(=O)=O

InChI

1S/Mo.2Na.2H2O.4O/h;;;2*1H2;;;;/q;2*+1;;;;;2*-1

InChI key

FDEIWTXVNPKYDL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

<ul>
<li><strong>Synthesis and characterization of Anderson-Evans type polyoxometalates, antibacterial properties:</strong> This study focuses on the synthesis and characterization of Anderson-Evans type polyoxometalates, utilizing Sodium molybdate dihydrate, exploring its potential in enhancing antibacterial properties (Avci Özbek H, 2023).</li>
<li><strong>Molybdenum Exposure in Drinking Water Vs Feed Impacts Apparent Absorption of Copper Differently in Beef Cattle Consuming a High-Forage Diet:</strong> This research examines how Sodium molybdate dihydrate, when introduced through water compared to feed, affects the absorption of copper in beef cattle, highlighting its role in agricultural nutrient management (Thorndyke MP et al., 2021).</li>
<li><strong>Molybdate Attenuates Lipid Accumulation in the Livers of Mice Fed a Diet Deficient in Methionine and Choline:</strong> This study demonstrates the effectiveness of Sodium molybdate dihydrate in reducing lipid accumulation in the livers of mice, offering insights into its potential therapeutic applications in liver health (Lee S et al., 2018).</li>
</ul>

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S W Ball et al.
Canadian journal of physiology and pharmacology, 80(3), 205-209 (2002-05-07)
A focus of current diabetes research is the development of insulinomimetic compounds for oral treatment of diabetes and its associated cardiac complications. Screening compounds for their potential insulinomimetic effects usually involves the use of radioactive isotopes. The focus of this
Saraswathi Panneerselvam et al.
Chemico-biological interactions, 145(2), 159-163 (2003-04-11)
Diabetes mellitus is a disease, which virtually affects all the systems in the body including the immune system. Bacterial infections are important causes of increased morbidity and mortality in diabetic patients. In the present study, we assessed the effect of
Akira Sakakura et al.
Chemical communications (Cambridge, England), (30)(30), 3561-3563 (2008-07-26)
Efficient total syntheses of fluvibactin and vibriobactin have been achieved via molybdenum(VI) oxide-catalyzed dehydrative cyclization, Sb(OEt)(3)-catalyzed ester-amide transformation, and WSCI and HOAt-promoted dehydrative amide formation.
Laurent Caron et al.
Journal of colloid and interface science, 282(2), 478-485 (2004-12-14)
Chemically generated singlet oxygen (1O2, 1Deltag) is able to oxidize a great deal of hydrophobic substrates from molybdate-catalyzed hydrogen peroxide decomposition, provided a suitable reaction medium such as a microemulsion system is used. However, high substrate concentrations or poorly reactive
Lluís Palou et al.
Pest management science, 58(5), 459-466 (2002-05-10)
The effectiveness of low-toxicity chemicals as possible alternatives to synthetic fungicides for the control of post-harvest green and blue moulds of citrus was evaluated. A preliminary selection of chemicals, mostly common food additives, was made through in vivo primary screenings

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service