Skip to Content
Merck
All Photos(2)

Key Documents

M1547

Sigma-Aldrich

Metronidazole

BioXtra

Synonym(s):

2-Methyl-5-nitroimidazole-1-ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9N3O3
CAS Number:
Molecular Weight:
171.15
Beilstein:
611683
EC Number:
MDL number:
UNSPSC Code:
51282808
PubChem Substance ID:
NACRES:
NA.76
Pricing and availability is not currently available.

product line

BioXtra

Quality Level

impurities

≤0.2% Total Impurities

ign. residue

≤0.1%

color

white to light yellow

mp

159-161 °C (lit.)

solubility

acetic acid: 0.1 M, clear, colorless to yellow

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
Na: ≤0.01%
Pb: ≤0.001%
Zn: ≤0.0005%

antibiotic activity spectrum

Gram-negative bacteria
parasites

Mode of action

DNA synthesis | interferes

storage temp.

2-8°C

SMILES string

CC1=NC=C([N+]([O-])=O)N1CCO

InChI

1S/C6H9N3O3/c1-5-7-4-6(9(11)12)8(5)2-3-10/h4,10H,2-3H2,1H3

InChI key

VAOCPAMSLUNLGC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: imidazole

Application

Metronidazole is used to study DNA damage and repair and has been found to induce the gene transfer agent VSH-1 in Brachyspira hyodysenteriae. It is commonly used for treatment of periodontitis and is used in bioavailability studies.

Biochem/physiol Actions

Metronidazole is a prodrug and is selective for anaerobic bacteria due to their ability to intracellularly reduce metronidazole to its active form. Reduced metronidazole covalently binds to DNA which disrupts its helical structure, induces DNA strand breaks and inhibits bacterial nucleic acid synthesis. Bacterial cell death results.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Muta. 1B - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tal Frolinger et al.
Scientific reports, 9(1), 3546-3546 (2019-03-07)
Dietary polyphenols promote memory in models of sleep deprivation (SD), stress, and neurodegeneration. The biological properties of dietary polyphenols greatly depend upon the bioavailability of their phenolic metabolites derivatives, which are modulated by gut microbiota. We recently demonstrated that supplementation
Fabrizio Palumbo et al.
Cell reports, 32(8), 108054-108054 (2020-08-28)
Operant learning requires multiple cognitive processes, such as learning, prediction of potential outcomes, and decision-making. It is less clear how interactions of these processes lead to the behavioral adaptations that allow animals to cope with a changing environment. We show
Christina M Surawicz et al.
The American journal of gastroenterology, 108(4), 478-498 (2013-02-27)
Clostridium difficile infection (CDI) is a leading cause of hospital-associated gastrointestinal illness and places a high burden on our health-care system. Patients with CDI typically have extended lengths-of-stay in hospitals, and CDI is a frequent cause of large hospital outbreaks
T H I Brummer et al.
BJOG : an international journal of obstetrics and gynaecology, 120(10), 1269-1276 (2013-06-22)
To evaluate cefuroxime and metronidazole antibiotic prophylaxis. Observational nonrandomised 1-year prospective cohort study. Fifty-three hospitals in Finland. A total of 5279 women undergoing hysterectomy for benign indications, with cefuroxime given to 4301 and metronidazole given to 2855. Excluding other antibiotics
Hitoshi Tsugawa et al.
PLoS biology, 18(9), e3000813-e3000813 (2020-09-30)
Short-chain fatty acids (SCFAs) produced by gastrointestinal microbiota regulate immune responses, but host molecular mechanisms remain unknown. Unbiased screening using SCFA-conjugated affinity nanobeads identified apoptosis-associated speck-like protein (ASC), an adaptor protein of inflammasome complex, as a noncanonical SCFA receptor besides

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service