Skip to Content
Merck
All Photos(1)

Documents

M1379

Sigma-Aldrich

Methyl α-D-galactopyranoside

≥99% (TLC)

Synonym(s):

Methyl α-D-galactoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14O6
CAS Number:
Molecular Weight:
194.18
Beilstein:
81570
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

bovine milk

Quality Level

Assay

≥99% (TLC)

form

powder or crystals

optical activity

[α]20/D 173 to 180°, c = 1.5% (w/v) in water

technique(s)

thin layer chromatography (TLC): suitable

color

white

mp

116-117 °C (lit.)

solubility

water: 50 mg/mL, clear, colorless

storage temp.

2-8°C

SMILES string

CO[C@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C7H14O6/c1-12-7-6(11)5(10)4(9)3(2-8)13-7/h3-11H,2H2,1H3/t3-,4+,5+,6-,7+/m1/s1

InChI key

HOVAGTYPODGVJG-PZRMXXKTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl α-D-galactopyranoside is potent inhibitor against the Debaryomyces hansenii UFV-1 extracellular and intracellular α-galactosidases.

Application

Methyl α-D-galactopyranoside has been used in computational studies of protonated β-d-galactose and its hydrated complex.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong-bin Xie et al.
The journal of physical chemistry. B, 116(16), 4851-4859 (2012-04-12)
We present an exploration of proton transfer dynamics in a monosaccharide, based upon ab initio molecular dynamic (AIMD) simulations, conducted "on-the-fly", in β-d-galactose-H(+) (βGal-H(+)) and its singly hydrated complex, βGal-H(+)-H2O. Prior structural calculations identify O6 as the preferred protonation site
Pollyanna A Viana et al.
Carbohydrate research, 346(5), 602-605 (2011-02-25)
α-D-Galactopyranosides were synthesized and their inhibitory activities toward the Debaryomyces hansenii UFV-1 extracellular and intracellular α-galactosidases were evaluated. Methyl α-D-galactopyranoside was the most potent inhibitor compared to the others tested, with K(i)(') values of 0.82 and 1.12 mmolL(-1), for extracellular
Alexander I Zinin et al.
Carbohydrate research, 337(7), 635-642 (2002-03-23)
1-O-Acetyl-beta-D-galactopyranose (AcGal), a new substrate for beta-galactosidase, was synthesized in a stereoselective manner by the trichloroacetimidate procedure. Kinetic parameters (K(M) and k(cat)) for the hydrolysis of 1-O-acetyl-beta-D-galactopyranose catalyzed by the beta-D-galactosidase from Penicillium sp. were compared with similar characteristics for
Satoru Takahashi et al.
Epilepsy research, 80(1), 18-22 (2008-05-06)
Glucose transporter 1 (GLUT1) deficiency syndrome is caused by a deficit in glucose transport to the brain during the pre- and postnatal periods. Here, we report two cases of GLUT1 deficiency syndrome diagnosed on the basis of clinical features, reduced
N Declerck et al.
Protein engineering, 7(8), 997-1004 (1994-08-01)
The L-arabinose binding protein (ABP) of Escherichia coli naturally binds L-arabinose and D-galactose with very high affinity and, with reduced affinity, a variety of other sugars that differ only at the C5 position of the pyranose ring. However, there are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service