Skip to Content
Merck

M0696

Anti-MNK2 (N-terminal) antibody produced in rabbit

enhanced validation

Enhanced Validation

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-GPRK7, Anti-MAP kinase interacting serine/threonine kinase 2, Anti-MKNK2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.44
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, IF, IP, WB
Citations:
6
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~50 kDa

species reactivity

rat (predicted), human, mouse (predicted)

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1.0 mg/mL

technique(s)

immunocytochemistry: 2-5 μg/mL using paraformadehyde fixed HEK-293T cells transfected with human MNK2, immunoprecipitation (IP): 2-4 μg using lysates of HEK-293T cells transfected with human MNK2, indirect immunofluorescence: suitable, western blot: 1-2 μg/mL using lysates of HEK-293T cells transfected with human MNK2

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MKNK2(2872)
mouse ... Mknk2(17347)
rat ... Mknk2(299618)

General description

MNK2 is a serine threonine kinase that phosphorylates endogenous eIF4E and thereby regulates protein translation and cell growth. This kinase inhibits apoptotic responses induced by arsenic trioxide. MNK2 has two distinct splice forms, namely; MNK2a and MNK2b. The localization and function of MNK2 splice variants are directed by their N and C termini . Anti-MNK2 (N-terminal) antibody is specific for human MNK2. The antibody is also expected to react with mouse and rat MNK2. In immunoblotting, staining of the MNK2 band is specifically inhibited by the immunizing peptide.

Immunogen

synthetic peptide corresponding to amino acids 51-64 of human MNK2 (corresponding to isoforms 1 and 2), conjugated to KLH via a C-terminal added cysteine residue. The corresponding sequence is conserved in human, rat, and mouse.

Application

Anti-MNK2 (N-terminal) antibody is suitable for use in immunocytochemistry (2-5 μg/mL using paraformaldehyde fixed, MNK2-transfected, HEK-293T cells) and indirect immunofluorescence. The antibody may also be used for immunoprecipitation (2-4 μg) and western blot (1-2 μg/mL) using lysates of HEK-293T cells transfected with human MNK2.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library