Skip to Content
Merck

M062200

Amino-ON CPG

pore size 1000 Å, 25-35μmol/g

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.51
UNSPSC Code:
41116105
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Proligo Reagents

pore size

1000 Å

storage temp.

−20°C

Legal Information

This product and it use is covered under US Patent Number 7,329,515


Still not finding the right product?

Explore all of our products under Amino-ON CPG


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library