M0567
Microsomes from Liver, Pooled
from human male
Synonym(s):
lab equipment accessory
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352204
NACRES:
NA.54
Recommended Products
biological source
human male
Quality Level
form
liquid
packaging
vial of ~10 mg
shipped in
dry ice
storage temp.
−70°C
Looking for similar products? Visit Product Comparison Guide
Application
Microsomes from Liver, Pooled has been used:
- in the glucuronidation kinetics assay to test the effects of herbal extracts on glucuronidation process
- as a human liver microsomes (HLM) matrix for testing metabolic stability of talazoparib using liquid chromatography-tandem mass spectrometry (LC–MS/MS)
- to study the metabolization of enantiomeric peptide D3
Microsomes from liver have been used in a study to assess differences in enzymatic activities between normal rat livers and from liver after partial hepatectomy. They have also been used in a study to investigate the carbon monoxide-binding pigment in liver microsomes.
Biochem/physiol Actions
Liver microsomes are subcellular particles derived from the endoplasmic reticulum of hepatic cells. These microsomes are a rich source of drug metabolizing enzymes, including cytochrome P-450. Microsome pools from various sources are useful in the study of xenobiotic metabolism and drug interactions.
N-glucuronidation of various 1-substituted imidazoles was found to occur in human liver microsomes.
Source of drug metabolizing enzymes, including cytochrome P-450.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Mohamed W Attwa et al.
Drug design, development and therapy, 14, 4439-4449 (2020-10-31)
Tandutinib (MLN518 or CT 53518) (TND) is a novel, oral, small-molecule inhibitor of type III receptor tyrosine kinases utilized for the treatment of acute myeloid leukemia (AML). In silico prediction of hepatic drug metabolism for TND was determined using the
THE CARBON MONOXIDE-BINDING PIGMENT OF LIVER MICROSOMES. I. EVIDENCE FOR ITS HEMOPROTEIN NATURE.
T OMURA et al.
The Journal of biological chemistry, 239, 2370-2378 (1964-07-01)
Sarvesh C Vashishtha et al.
Drug metabolism and disposition: the biological fate of chemicals, 30(10), 1070-1076 (2002-09-14)
N-Glucuronidation at an aromatic tertiary amine of 5-membered polyaza ring systems was investigated for a model series of eight 1-substituted imidazoles in liver microsomes from five species. The major objectives were to investigate substrate specificities of the series in human
Anne Elfgen et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 107, 203-207 (2017-07-18)
The aggregation of the amyloid β protein (Aβ) plays an important role in the pathology of Alzheimer's disease. Previously, we have developed the all-d-enantiomeric peptide D3, which is able to eliminate neurotoxic Aβ oligomers in vitro and improve cognition in
Ali S Abdelhameed et al.
Royal Society open science, 6(6), 190434-190434 (2019-07-18)
Spebrutinib (SBT) is a Bruton's tyrosine kinase inhibitor. SBT is currently in phase II and phase I clinical trials for the management of rheumatoid arthritis and chronic lymphocytic leukaemia, respectively. We developed and validated a liquid chromatography tandem mass spectrometry
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service