Skip to Content
Merck

Skip To

LSMLS06

LSMLS Plate 6 (Lipophilic)

Supplied by IROA Technologies

Synonym(s):

LSMLS, Large Scale Metabolite Libarary of Standards, Metabolic Library, Metabolite Library

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41116107
NACRES:
NA.21
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solid

packaging

pkg of 1 plates

manufacturer/tradename

IROA Technologies

plate size

96 wells

shipped in

dry ice

storage temp.

−20°C

General description

LSMLS Plate 6 is a collection of high quality small biochemical molecules that span a broad range of primary metabolism. These are high purity (>95%) compounds supplied in an economical, ready-to-use format. This library of standards is most commonly used for metabolite qualification and quantification for MS and NMR, functional cellular assays, and phenotypic screening. Metabolite descriptors provided with product include: Name, Parent CID, molecular formula, molecular weight, CAS, ChEBI, HMDB ID, PubChem Compound, and Substance ID.The library is intended to be used for mass spectrometry metabolomics applications and provides a broad representation of primary metabolism.

Application

Metabolite qualification and quantification for MS, NMR, etc.; Functional cellular assays; Phenotypic screening. Suitable for manual and automated work flows.

Features and Benefits

To view a complete list of compounds in the library for recent lots, download the plate maps below:
The complete plate map with name, Parent CID, molecular formula, molecular weight, CAS, ChEBI, HMBD ID, PubChem Compound, and Substance ID are provided with the software download.

Broad metabolite spectrum, key primary metabolites and intermediates covering key metabolic pathways, including the following classes of compounds:
  • Carboxylic acids, amino acids
  • Biogenic amines, polyamines
  • Nucleotides, coenzymes and vitamins
  • Mono- and disaccharides
  • Fatty acids, lipids, steroids and hormones

Preparation Note

LSMLS Plate 6 compounds are conveniently provided at 1 mg per well, enough for multiple injections. Compounds can either be used as standards and injected individually or a strategy may be employed to pool compounds by row to allow multiple mixtures to be analyzed per injection. For more information on LSMLS06, solubilization and preparation notes please refer to the product information sheet.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Legal Information

LSMLS is a trademark of IROA Technologies LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MSMLS06LSMLS07MSMLS07
description

Supplied by IROA Technologies

description

Supplied by IROA Technologies

description

Supplied by IROA Technologies

description

Supplied by IROA Technologies

manufacturer/tradename

IROA Technologies

manufacturer/tradename

IROA Technologies

manufacturer/tradename

IROA Technologies

manufacturer/tradename

IROA Technologies

form

solid

form

solid

form

liquid

form

solid

packaging

pkg of 1 plates

packaging

pkg of 1 plates

packaging

pkg of 1 plates

packaging

pkg of 1 plates

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

plate size

96 wells

plate size

96 wells

plate size

96 wells

plate size

96 wells

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Muta. 2 - Repr. 2 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2 Inhalation - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters