Skip to Content
Merck

L7414

N-Lauroylsarcosine sodium salt solution

20%, Molecular Biology

Synonym(s):

Sarcosyl, Sarkosyl NL, Sodium N-dodecanoyl-N-methylglycinate, Sodium N-lauroylsarcosinate solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3(CH2)10CON(CH3)CH2COONa
CAS Number:
Molecular Weight:
293.38
UNSPSC Code:
12161900
NACRES:
NA.25
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
5322974
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

plant

Quality Segment

grade

Molecular Biology

description

anionic

assay

19-21%

form

liquid

mol wt

293.38 g/mol

concentration

19-21% (Titration by HClO4), 20%

solubility

water: soluble at 20 °C

foreign activity

DNase, RNase, and protease, none detected

SMILES string

[Na+].CCCCCCCCCCCC(=O)N(C)CC([O-])=O

InChI

1S/C15H29NO3.Na/c1-3-4-5-6-7-8-9-10-11-12-14(17)16(2)13-15(18)19;/h3-13H2,1-2H3,(H,18,19);/q;+1/p-1

InChI key

KSAVQLQVUXSOCR-UHFFFAOYSA-M

General description

N-Lauroylsarcosine sodium salt solution is an anionic surfactant which can be used in DNA extraction.

Application

N-Lauroylsarcosine sodium salt has been used in a study to assess the mechanisms underlying surfactant-induced itching. It has also been used in a study to investigate the formation of pH-responsive, stable vesicles resulting from an interaction with N-cetylpyridinium chloride (CPC) and N-dodecylpyridinium chloride (DPC) in aqueous mixtures.

Features and Benefits

N-Lauroylsarcosine does not precipitate and is commonly used in place of SDS.


Still not finding the right product?


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

512.6 °F

flash_point_c

267 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library