Skip to Content
Merck

L5380

Lectin from Pisum sativum (pea)

lyophilized powder

Synonym(s):

Pisum sativum agglutinin, PSA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.32
UNSPSC Code:
12352202
Form:
lyophilized powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

lyophilized powder

Quality Segment

potency

<20 μg/mL per mL agglutination activity

composition

Protein, ≥70% biuret

storage temp.

2-8°C

Biochem/physiol Actions

PSA is not blood group specific, but has an affinity for terminal α-D-mannosyl residues. PSA is a mitogen similar to concanavalin A.

Preparation Note

Conjugates are prepared from affinity purified lectin.
Purified by affinity chromatography.

Analysis Note

Agglutination activity is expressed in μg/ml and is determined from serial dilutions in phosphate buffered saline, pH 6.8, of a 1 mg/ml solution. This activity is the lowest concentration to agglutinate a 2% suspension of human erythrocytes after 1 hr incubation at 25 °C.


Still not finding the right product?

Explore all of our products under Lectin from Pisum sativum (pea)


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library