Skip to Content
Merck
All Photos(1)

Key Documents

L4897

Sigma-Aldrich

Luteinizing hormone releasing hormone salmon

≥97% (HPLC)

Synonym(s):

GnRH, Gonadoliberin, Gonadotropin releasing hormone, LH-RH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C60H73N15O13
CAS Number:
Molecular Weight:
1212.31
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.32
Pricing and availability is not currently available.

Quality Level

Assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CC(C)CC(NC(=O)C(Cc1c[nH]c2ccccc12)NC(=O)CNC(=O)C(Cc3ccc(O)cc3)NC(=O)C(CO)NC(=O)C(Cc4c[nH]c5ccccc45)NC(=O)C(Cc6cnc[nH]6)NC(=O)C7CCC(=O)N7)C(=O)N8CCCC8C(=O)NCC(N)=O

InChI

1S/C60H73N15O13/c1-32(2)20-47(60(88)75-19-7-12-49(75)59(87)65-28-50(61)78)73-55(83)44(22-34-25-63-40-10-5-3-8-38(34)40)69-52(80)29-66-53(81)43(21-33-13-15-37(77)16-14-33)70-58(86)48(30-76)74-56(84)45(23-35-26-64-41-11-6-4-9-39(35)41)71-57(85)46(24-36-27-62-31-67-36)72-54(82)42-17-18-51(79)68-42/h3-6,8-11,13-16,25-27,31-32,42-49,63-64,76-77H,7,12,17-24,28-30H2,1-2H3,(H2,61,78)(H,62,67)(H,65,87)(H,66,81)(H,68,79)(H,69,80)(H,70,86)(H,71,85)(H,72,82)(H,73,83)(H,74,84)

InChI key

NMJREATYWWNIKX-UHFFFAOYSA-N

Related Categories

Amino Acid Sequence

Glp-His-Trp-Ser-Tyr-Gly-Trp-Leu-Pro-Gly-NH2

Application

Luteinizing hormone releasing hormone (salmon) has been used to study the effect of LHRH (luteinizing hormone releasing hormone) in the brain of Podarcis sicula sicula (male) during reproduction.[1] It has been used to study the hormonal regulation of PACAP (pituitary adenylate cyclase-activating polypeptide) and its receptor, PAC1 (adenylate cyclase activating polypeptide 1 (pituitary) receptor type I).[2]

Biochem/physiol Actions

Hypothalamic peptide that stimulates release of gonadotrophins from anterior pituitary, thus regulating reproductive functions.
Stimulates release of gonadotrophins from anterior pituitary.

Other Notes

Lyophilized from 0.5% acetic acid

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vasoactive Intestinal Peptide Indirectly Elicits Pituitary LH Secretion Independent of GnRH in Female Zebrafish.
Tanaka, et al.
Endocrinology, 163 (2022)
Weiming Zheng et al.
Reproductive sciences (Thousand Oaks, Calif.), 21(1), 41-51 (2013-05-22)
Pituitary adenylate cyclase-activating polypeptide (PACAP) and its receptors are expressed in the hypothalamus, the gonadotrope cells of the anterior pituitary gland, and the gonads, forming an autocrine-paracrine system in these tissues. Within the pituitary, PACAP functions either alone or synergistically
A Gobbetti et al.
Journal of reproduction and fertility, 101(3), 523-529 (1994-08-01)
The release of PGF2 alpha and PGE2, progesterone, androgens and oestradiol in vitro, and the aromatase activity in the brain of the male lizard Podarcis sicula sicula during three different phases of the reproductive period were evaluated. In addition, the
Shel-Hwa Yeo et al.
Neuroendocrinology, 100(2-3), 191-197 (2014-10-11)
Kisspeptin-Gpr54 signaling is critical for regulating the activity of gonadotropin-releasing hormone (GnRH) neurons in mammals. Previous studies have shown that the negative feedback mechanism is disrupted in global Gpr54-null mutants. The present investigation aimed to determine (1) if a lack
Araceli Diez-Fraile et al.
Human fertility (Cambridge, England), 17(2), 90-98 (2014-04-02)
Reproductive life span and fertility have been shown to depend on successful early folliculogenesis, which involves cell-to-cell communication and the concerted regulation of gene expression at both the oocyte and granulosa cell levels. Recently, micro RNAs (miRNAs) were identified as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service