Skip to Content
Merck
All Photos(2)

Key Documents

L4894

Sigma-Aldrich

Lactoferrin from human milk

powder, BioReagent, suitable for cell culture

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.75
Pricing and availability is not currently available.

product line

BioReagent

Quality Level

Assay

≥85% (SDS-PAGE)

form

powder

mol wt

82.4 kDa

technique(s)

cell culture | mammalian: suitable

color

white to light red

solubility

PBS, pH 7.4: 1 mg/mL, clear to slightly hazy (0.01 M phosphate buffer, 0.0027 M potassium chloride and 0.137 M sodium chloride, pH 7.4, at 25 °C)

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

SMILES string

N([C@@H](C(C)C)C(=O)N[C@@H](C)C(=O)N[C@@H](Cc1ccccc1)C(=O)N[C@@H](C(C)C)C(=O)O)C(=O)[C@@H](NC(=O)CNC(=O)[C@@H](NC(=O)[C@@H](N)CC(=O)N)C)CC(=O)O

InChI

1S/C35H53N9O12/c1-16(2)27(43-33(53)23(14-26(47)48)41-25(46)15-38-29(49)18(5)39-31(51)21(36)13-24(37)45)34(54)40-19(6)30(50)42-22(12-20-10-8-7-9-11-20)32(52)44-28(17(3)4)35(55)56/h7-11,16-19,21-23,27-28H,12-15,36H2,1-6H3,(H2,37,45)(H,38,49)(H,39,51)(H,40,54)(H,41,46)(H,42,50)(H,43,53)(H,44,52)(H,47,48)(H,55,56)/t18-,19-,21-,22-,23-,27-,28-/m0/s1

InChI key

QCBUWCQOKPLTDZ-PKRULZLPSA-N

Gene Information

human ... LTF(4057)

Looking for similar products? Visit Product Comparison Guide

General description

Lactoferrin is mapped to human chromosome 3p21.31. It comprises globular lobes and undergoes post-translational modifications like phosphorylation and glycosylation.[1] Lactoferrin belongs to the lactotransferrin family and has Ser‐Lys catalytic dyad like serine proteases.[2]

Application

Lactoferrin from human milk has been used:
  • to test its neuroprotective effect on neuroblastoma cells and mouse mesencephalic dopaminergic cell lines MN9D [3]
  • to test its antioxidative effect in Edible Bird′s Nest (EBN) induced toxicity in human neuroblastoma SH-SY5Y cells using oxygen radical absorbance capacity (ORAC) assay and 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) assay[4]
  • in the preparation of immobilized titanium substrate for testingMG-63 osteoblast activity[5]
  • as a low-density lipoprotein (LDL) receptor-related protein (LRP1) ligand in vascular smooth muscle cells (VSMC)[6]

Biochem/physiol Actions

Lactoferrin is an iron binding protein. It is structurally similar to transferrin, the plasma iron transport protein; but lactoferrin has a much higher affinity for iron (250 fold). It is very abundant in colostrum and small amounts can also be found in tears, saliva, mucous secretions and in the secondary granules of neutrophils. It is made by mucosal epithelium and neutrophils and is released by these cells in response to inflammatory stimuli. Bacterial growth is inhibited by its ability to sequester iron and also permeabilize bacterial cell walls by binding to lipopolysaccharides through its N-terminus. Lactoferrin can inhibit viral infection by binding tightly to the viral envelope protein. This prevents cell-virus fusion by blocking the binding domain. Lactoferrin appears to activate host defense systems in part by stimulating the release of interleukin-8, a neutrophil activator. It may also be involved in antibody and interleukin synthesis, lymphocyte proliferation and complement activation.
Lactoferrin is blood-brain barrier (BBB) traversable and has therapeutic potential to regulate inflammation, immunity and apoptosis in Parkinson′s disease.[1] It promotes the expression of brain-derived neurotrophic factor (BDNF) via various signaling pathways and aids protection in motor dysfunction.[1] Lactoferrin favors osteogenic differentiation and proliferation of human adipose-derived stem cells (hADSCs).[3] It exhibits proteolytic activity on Haemophilus influenzae colonization factors.[2] Lactoferrin is also a nutraceutical and a potential marker in the pathophysiology of inflammatory bowel diseases, leukocytes[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Bellamy et al.
Biochimica et biophysica acta, 1121(1-2), 130-136 (1992-05-22)
We report the existence of a previously unknown antimicrobial domain near the N-terminus of lactoferrin in a region distinct from its iron-binding sites. A single active peptide representing this domain was isolated following gastric pepsin cleavage of human lactoferrin, and
Linyuan Ma et al.
Scientific reports, 8(1), 7433-7433 (2018-05-11)
Efficient transgene expression in recipient cells constitutes the primary step in gene therapy. However, random integration in host genome comprises too many uncertainties. Our study presents a strategy combining bioinformatics and functional verification to find transgene integration sites in pig
Chuang Guo et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 42(13), 2504-2515 (2017-01-13)
Growing evidence suggests that lactoferrin (Lf), an iron-binding glycoprotein, is a pleiotropic functional nutrient. In addition, Lf was recently implicated as a neuroprotective agent. These properties make Lf a valuable therapeutic candidate for the treatment of Alzheimer's disease (AD). However
José M Fernández-Real et al.
Obesity (Silver Spring, Md.), 18(3), 482-488 (2009-08-22)
Lactoferrin is an innate immune system protein with anti-inflammatory and antioxidant activities. We aimed to evaluate circulating lactoferrin levels in association with lipid concentrations, and parameters of oxidative stress and inflammation in subjects with morbid obesity after an acute fat
Lactoferrin: a multifunctional immunoregulatory protein?
J Brock
Immunology today, 16(9), 417-419 (1995-09-01)

Questions

  1. Could you confirm whether products SRP6519 and L4894 Lactoferrin derived from human milk have confirmed iron content?

    1 answer
    1. The specification sheets for products SRP and L4894 do not mention testing for iron. The product SRP6519 undergoes basic testing, including appearance and SDS Page electrophoresis. On the other hand, product L4894 undergoes more detailed testing, including appearance, solubility, protein by biuret, SDS Page electrophoresis, as well as additional testing for infectious agents and use in cell culture. While iron is expected to be present, there are no confirmatory tests performed to document the amount of iron on a batch-to-batch basis.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service